AXYFX384R
Pipette tips for Beckman® FX Robotics System
racked, capacity 30 μL, pkg of 5x3840tips/cs, clear
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
30 μL
sterility:
non-sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc FX-384-R
Corning
Corning
material
clear
sterility
non-sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
manufacturer/tradename
Axygen Inc FX-384-R
Corning
packaging
pkg of 5x3840tips/cs
capacity
30 μL
volume
30 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Legal Information
Beckman is a registered trademark of Beckman Coulter, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service