Skip to Content
Merck
All Photos(4)

Key Documents

AV49134

Sigma-Aldrich

Anti-SULT1A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HAST1/HAST2, Anti-MGC131921, Anti-MGC5163, Anti-P-PST, Anti-PST, Anti-ST1A3, Anti-STP, Anti-Sulfotransferase family, cytosolic, 1A, phenol-preferRing, member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

human, horse, rabbit, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SULT1A1(6817)

Immunogen

Synthetic peptide directed towards the N terminal region of human SULT1A1

Application

Anti-SULT1A1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1 (SULT1A1) is a phenol sulfotransferase with thermostable activity. The members of sulfotransferase family localize to cytoplasm and catalyze the sulfate conjugation of hormones, xenobiotic compounds, drugs and neurotransmitters.

Sequence

Synthetic peptide located within the following region: ELIQDTSRPPLEYVKGVPLIKYFAEALGPLQSFQARPDDLLISTYPKSGT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Hebbring et al.
Cytogenetic and genome research, 123(1-4), 205-210 (2008-01-01)
Pharmacogenetics is the study of the role of inheritance in variation to drug response. Drug response phenotypes can vary from adverse drug reactions at one end of the spectrum to equally serious lack of the desired effect of drug therapy
Philip M Probert et al.
Toxicology letters, 243, 98-110 (2016-01-08)
Rat B-13 progenitor cells are readily converted into functional hepatocyte-like B-13/H cells capable of phase I cytochrome P450-dependent activation of pro-carcinogens and induction of DNA damage. The aim of the present study was to investigate whether the cells are also
M W H Coughtrie
The pharmacogenomics journal, 2(5), 297-308 (2002-11-20)
Members of the cytosolic sulfotransferase (SULT) superfamily catalyse the sulfation of a multitude of xenobiotics, hormones and neurotransmitters. Humans have at least 10 functional SULT genes, and a number of recent advances reviewed here have furthered our understanding of SULT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service