Skip to Content
Merck
All Photos(1)

Key Documents

AV47036

Sigma-Aldrich

Anti-TOR1B antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DQ1, Anti-MGC4386, Anti-Torsin family 1, member B (torsin B)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
rat, mouse, human
technique(s):
western blot: suitable
citations:
3

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TOR1B(27348)

Immunogen

Synthetic peptide directed towards the C terminal region of human TOR1B

Application

Anti-TOR1B antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/mL.

Biochem/physiol Actions

TOR1B [Torsin family 1, member B (torsin B)] gene encodes a 336 amino acid containing protein, predominantly expressed in the developing human brain and belonging to ClpA/ClpB family and torsin subfamily. It plays a pivotal role in postnatal developmental events and is crucial for proper neurological development. TOR1B may also facilitate as a molecular chaperone assisting in the proper folding of secreted and membrane proteins. Mutation in TOR1B gene results in torsion dystonia, an autosomal dominant neurological disorder.

Sequence

Synthetic peptide located within the following region: VFNNKHSGLWHSGLIDKNLIDYFIPFLPLEYRHVKMCVRAEMRARGSAID

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anju Vasudevan et al.
Brain research, 1073-1074, 139-145 (2006-02-07)
Early onset torsion dystonia is characterized by involuntary movements and distorted postures and is usually caused by a 3-bp (GAG) deletion in the DYT1 (TOR1A) gene. DYT1 codes for torsinA, a member of the AAA+ family of proteins, implicated in
E Bahn et al.
Brain research, 1116(1), 112-119 (2006-08-30)
Familial, early onset, generalized torsion dystonia is the most common and severe primary dystonia. The majority of cases are caused by a 3-bp deletion (GAG) in the coding region of the DYT1 (TOR1A) gene. The cellular and regional distribution of
C Kamm et al.
Neurology, 67(10), 1857-1859 (2006-11-30)
Recently, association of a TOR1A(DYT1)/TOR1B risk haplotype with common forms of idiopathic dystonia has been reported in the Icelandic population. Here we report a strong association of two single nucleotide polymorphisms within or in close proximity to the TOR1A 3'UTR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service