Skip to Content
Merck
All Photos(12)

Key Documents

AMAB91033

Sigma-Aldrich

Anti-Glial Fibrillary Acidic Protein Antibody

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, mouse monoclonal, CL2713

Synonym(s):

FLJ45472

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

Product Name

Monoclonal Anti-GFAP antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2713, purified immunoglobulin, buffered aqueous glycerol solution

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CL2713, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

orthogonal RNAseq
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 1 μg/mL
immunohistochemistry: 1:5000- 1:10000

isotype

IgG1

immunogen sequence

LEGEENRITIPVQTFSNLQIRETSLDTKSVSEGHLKRNIVVKTVEMRDGEVIKESKQEHKD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GFAP(2670)

Immunogen

glial fibrillary acidic protein

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76154

Physical form

40% glycerol and PBS (pH 7.2). 0.02% sodium azide is added as preservative.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lianyan Huang et al.
Frontiers in neural circuits, 14, 581435-581435 (2020-12-17)
Aging is accompanied by a progressive decrease in learning and memory function. Synaptic loss, one of the hallmarks of normal aging, likely plays an important role in age-related cognitive decline. But little is known about the impact of advanced age
Mickaël Soued et al.
BJA open, 3, 100029-100029 (2023-08-17)
The p38 protein is a ubiquitous mitogen-activated protein kinase involved in the proinflammatory signalling pathway and in the pain response after various noxious stimuli. Many p38 inhibitors have been developed and shown to provide effective analgesia in animal models. They
Igor Manzhulo et al.
Brain sciences, 11(12) (2021-12-25)
The present study demonstrates that synaptamide (N-docosahexaenoylethanolamine), an endogenous metabolite of docosahexaenoic acid, when administered subcutaneously (4 mg/kg/day, 14 days), exhibits analgesic activity and promotes cognitive recovery in the rat sciatic nerve chronic constriction injury (CCI) model. We analyzed the
Yingjun Liu et al.
Scientific reports, 8(1), 14600-14600 (2018-10-04)
Prion diseases, Alzheimer's disease and Parkinson's disease (PD) are fatal degenerative disorders that share common neuropathological and biochemical features, including the aggregation of pathological protein conformers. Lymphocyte activation gene 3 (Lag3, also known as CD223) is a member of the
Anna Starinets et al.
Cells, tissues, organs, 211(1), 73-84 (2021-09-13)
At present, there is a growing interest in the study of the neurotropic activity of polyunsaturated fatty acid ethanolamides (N-acylethanolamines). N-docosahexaenoylethanolamine (DHEA), or synaptamide, an endogenous metabolite of docosahexaenoic acid, is a promising compound with anti-inflammatory activity. The results of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service