Skip to Content
Merck
All Photos(1)

Key Documents

ADD620C

Sigma-Aldrich

HepaRG Maintenance and Metabolism Medium Supplement with Antibiotics

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

shipped in

dry ice

storage temp.

−20°C

General description

Biopredic′s HepaRG Maintenance and Metabolism Medium Supplement with Antibiotics can be used with all HepaRG cell lines. See user guide for detailed protocols.
HepaRG cells uniquely maintain significant levels of hepatic cell functions, being CYP450 inducible and support the complete hepatitis B virus (HBV) replicative cycle.

Application

HepaRG maintenance and metabolism medium supplement with antibiotics has been used to culture HepaRG cells:
  • to study the interaction of hepatocytes and cholangiocytes when challenged with chlorpromazine (CPZ)
  • to assess the validity of stable reference genes (RG) for gene expression using quantitative real-time polymerase chain reaction (RT-qPCR)
  • as a supplement in basal hepatic cell culture medium to study the effects of cholestasis on liver connexins

Legal Information

Use of this product is subject to one or more license agreements.

HepaRG cells are patented and their use is strictly limited; consider the cells as a single use, disposable product that must be destroyed upon conclusion of a study or experiment. Propagating, reproducing, cloning, subcloning or any other use of the cells following the conclusion of a study is prohibited. Use of the cells to produce or manufacture commercial products for general sale or for use in the manufacture of products intended for general sale is prohibited. Transfer of the cells to anyone not employed within the same organization, whether for financial benefit or not, is prohibited. If you are unwilling to accept the terms of this LIMITED USE LICENSE, do not ORDER or use them and immediately return the cells for credit. Violators of this Limited Use Licesense will be prosecuted to the fullest extent of the law.
HepaRG is a trademark of BioPredic International company

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katie Morgan et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 111, 1408-1416 (2019-03-08)
Chlorpromazine (CPZ) is a neuroleptic drug and prototype compound used to study intrahepatic cholestasis. The exact mechanisms of CPZ induced cholestasis remain unclear. Rat hepatocytes, or a sandwich culture of rat and human hepatocytes, have been the most commonly used
Axelle Cooreman et al.
International journal of molecular sciences, 21(18) (2020-09-11)
Connexins are goal keepers of tissue homeostasis, including in the liver. As a result, they are frequently involved in disease. The current study was set up to investigate the effects of cholestatic disease on the production of connexin26, connexin32 and
Joanna Brzeszczyńska et al.
Cells, 9(3) (2020-04-05)
Gene expression analysis by quantitative real-time polymerase chain reaction (RT-qPCR) is routinely used in biomedical studies. The reproducibility and reliability of the data fundamentally depends on experimental design and data interpretation. Despite the wide application of this assay, there is
Anika Mann et al.
Human cell, 30(4), 267-278 (2017-05-21)
HepaRG cells are widely used as an in vitro model to assess drug-induced hepatotoxicity. However, only few studies exist so far regarding their suitability to detect the effects of drugs requiring a preceding activation via the cytochrome P450 (CYP) system.
Yi Ni et al.
Methods in molecular biology (Clifton, N.J.), 1540, 15-25 (2016-12-16)
Investigations of virus-host interactions rely on suitable in vitro cell culture systems that efficiently support virus infection. Such systems should ideally provide conditions that resemble those of natural host cells, e.g., the cell-type specific signaling and metabolic pathways. For HBV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service