Skip to Content
Merck
All Photos(1)

Documents

ABK0010

Sigma-Aldrich

EpoBridge ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

antibody form

IgG fraction of antiserum

species reactivity

human (Human IgG)

shipped in

wet ice

storage temp.

2-8°C

Application

For research use only. Not for use in diagnostic procedures.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xing Wang et al.
Frontiers in pharmacology, 4, 103-103 (2013-08-24)
Antibody arrays were developed to probe a monoclonal antibody's three-dimensional structure (3-D structure). Peptides with overlapping regions were designed to cover the whole mAb light chain and heavy chain, respectively, and used to generate polyclonal antibodies after the conjugation of
Biosimilar mAb in-process Sample Higher Order Structure Analysis with Protein Conformational Array ELISA.
Michael Davies, Gan Wang, Jian Gong, Guofeng Fu and Xing Wang
Pharmaceut. Res., 1-11 (2016)
Yuanli Song et al.
mAbs, 10(3), 397-405 (2018-01-10)
The elucidation of antibody higher order structure (HOS) is critical in therapeutic antibody development. Since HOS determines the protein bioactivity and chemo-physical properties, this knowledge can help to ensure that the safety and efficacy attributes are not compromised. Protein conformational
Soon Kwan Jung et al.
mAbs, 6(5), 1163-1177 (2014-12-18)
Remsima (infliximab) was recently approved as the world's first biosimilar monoclonal antibody (mAb) in both the European Union and Korea. To achieve this, extensive physicochemical characterization of Remsima in relation to Remicade was conducted in order to demonstrate the highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service