AA0100
Ammonia Assay Kit
sufficient for 100 assays
Synonym(s):
Ammonia Quantitative Test Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
sufficient for 100 assays
storage temp.
2-8°C
Application
Ammonia concentration was measured in serum and urine using the Sigma Ammonia Assay Kit in a study assessing expression of renal ammonia transporters in response to chromic metabolic acidosis.
This kit is for the quantitative, enzymatic determination of ammonia in food and biological samples. Ammonia reacts with α-ketoglutaric acid (KGA) and reduced nicotinamide adenine dinucleotide phosphate (NADPH) in the presence of L-glutamate dehydrogenase (GlDH) to form L-glutamate and oxidized nicotinamide adenine dinucleotide phosphate (NADP+). The decrease in absorbance at 340 nm, due to the oxidation of NADPH, is proportional to the ammonia concentration. L-Glutamate dehydrogenase reacts specifically with ammonia. The Ammonia Assay Kit may be used to determine ammonia concentrations in the range of 0.2-15 μg/ml.
Kit Components Only
Product No.
Description
- Ammonia Assay Reagent 10 vial(s)
- L-Glutamate Dehydrogenase for Ammonia Assay 1 mL/vial
- Ammonia Standard Solution 1 mL/vial
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Bioprocess and biosystems engineering, 42(5), 711-725 (2019-01-24)
Mammalian-inducible expression systems are increasingly available and offer an attractive platform for the production of recombinant proteins. In this work, we have conducted process development for a cumate-inducible GS-CHO cell-line-expressing rituximab. To cope with the limitations encountered in batch when
Scientific reports, 7(1), 9510-9510 (2017-08-27)
We combined
Fish & shellfish immunology, 65, 226-234 (2017-04-22)
Ammonia is highly toxic to aquatic organisms, but whether ammonia excretion or ammonia metabolism to less toxic compounds is the major strategy for detoxification in marine fish against chronic ammonia exposure is unclear to date. In this study, we investigated
Nature communications, 11(1), 108-108 (2020-01-09)
In cnidarian-Symbiodiniaceae symbioses, algal endosymbiont population control within the host is needed to sustain a symbiotic relationship. However, the molecular mechanisms that underlie such population control are unclear. Here we show that a cnidarian host uses nitrogen limitation as a
BMC plant biology, 19(1), 239-239 (2019-06-07)
Ammonium transporters (AMTs), a family of proteins transporting ammonium salt and its analogues, have been studied in many aspects. Although numerous studies have found that ammonium affects the interaction between plants and pathogens, the role of AMTs remains largely unknown
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service