Skip to Content
Merck
All Photos(1)

Key Documents

A6608

Sigma-Aldrich

Albumin human

recombinant, expressed in Saccharomyces cerevisiae, aqueous solution, 10% in aqueous buffer, ≥99% (agarose gel electrophoresis)

Synonym(s):

rHSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2936H4624N786O889S41
CAS Number:
Molecular Weight:
66471.53
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.25

biological source

human

Quality Level

recombinant

expressed in Saccharomyces cerevisiae

Assay

≥99% (agarose gel electrophoresis)

form

aqueous solution

mol wt

monomer ~67 kDa

concentration

10% in aqueous buffer

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin constitutes 25% of the proteins produced by the liver and is a prominent protein found in the blood. It is a protein of a single polypeptide chain with a thiol group.

Application

Albumin human has been used:
  • to study its adsorption process on model mica surface at pH 3.5 using the atomic force microscopy (AFM) and in situ streaming potential measurements
  • to study its proteolytic activities under reducing conditions
  • in the medium for stem cell cultivation

This recombinant variant is recommended for use in absorption, distribution, metabolism, and excretion (ADME) pharmacological research; cell culture; drug delivery research; and cryopreservation of cells.

Biochem/physiol Actions

Albumin is a binding and transport protein that serves as a protein repository and a carrier for amino acids. Albumin binding dissolves the biomolecules in blood and ensures its effective transport. This also prevents the loss of substance through excretion. Pathological conditions like vasculitis, lymphangiectasia, glomerulonephritis, and hepatic sinusoidal hypertension might result in hypoalbuminemia. This product can be used in the pharmaceutical industry as a drug excipient and to coat medical devices.
Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.

Analysis Note

Endotoxin tested

Other Notes

View more information on human serum albumin.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitry Kryndushkin et al.
Pharmaceutical research, 33(2), 526-539 (2015-10-27)
Ascorbic acid has been considered as a potential radical scavenging excipient for pharmaceutical formulations. However, under certain circumstances, ascorbic acid can generate reactive oxygen species via redox cycling. The objective of this study was to investigate ascorbic acid-induced oxidative carbonylation
Dmitry Kryndushkin et al.
Pharmaceutical research, 34(4), 765-779 (2017-02-06)
Protein carbonylation is an irreversible modification of Lys, Arg, Thr and Pro amino acids under conditions of oxidative stress. Previous studies have reported specific carbonylated residues in purified recombinant albumins, albeit with a lack of agreement between the studies. Currently
Serum proteins and the dysproteinemias
Clinical biochemistry of domestic animals, 117-138 (1997)
Veronica Liv Andersen et al.
Theranostics, 9(9), 2662-2677 (2019-05-28)
Rationale: Within the field of personalized medicine there is an increasing focus on designing flexible, multifunctional drug delivery systems that combine high efficacy with minimal side effects, by tailoring treatment to the individual. Methods: We synthesized a chemically stabilized ~4
Marta Kujda et al.
Colloids and surfaces. B, Biointerfaces, 127, 192-199 (2015-02-14)
Recombinant human serum albumin (rHSA) in monomeric state is widely used in pharmaceutical industry as a drug excipient and for preparing coatings for medical devices. In this work the adsorption process of rHSA on model mica surface at pH 3.5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service