Skip to Content
Merck
All Photos(3)

Documents

A6103

Sigma-Aldrich

Aprotinin

recombinant, expressed in Nicotiana (tobacco), ≥5 TIU/mg protein, powder, ≥98% (SDS-PAGE), suitable for inhibition assay

Synonym(s):

BPTI, Basic pancreatic trypsin inhibitor, Kallikrein-trypsin inactivator, Kunitz protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Aprotinin Nicotiana tobacco, >= 5TIU/mg protein, >= 98 % SDS-PAGE | 9087-70-1, recombinant, expressed in Nicotiana (tobacco), ≥5 TIU/mg protein, ≥98% (SDS-PAGE)

recombinant

expressed in Nicotiana (tobacco)

description

recombinant form of thenative bovine-sequence aprotinin purified from plant tissue (tobacco)

product line

BioUltra

Assay

≥98% (SDS-PAGE)

form

powder

specific activity

≥5 TIU/mg protein

mol wt

~_6.5 kDa

technique(s)

inhibition assay: suitable

solubility

0.85% sodium chloride: 5 mg/mL

UniProt accession no.

storage temp.

2-8°C

Gene Information

cow ... PTI(404172)

Unit Definition

One trypsin inhibitor unit (TIU) will decrease the activity of 2 trypsin units by 50% where one trypsin unit will hydrolyze 1.0 μmole of N-α-benzoyl-DL-arginine p-nitroanilide (BAPNA) per min at pH 7.8 at 25 °C.

Preparation Note

Contains no animal-derived components or impurities, and is manufactured by transient expression of the aprotinin message in RNA (+)-strand tobacco mosaic virus vectors propagated in non-transgenic Nicotiana plants. This is a recombinant form of the native, bovine-sequence aprotinin, which is traditionally isolated from bovine lung by methods involving fractional precipitation, gel filtration, and ion exchange chromatography. Unlike animal-derived aprotinin, this product is isolated and purified from plant tissue by proprietary methods.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allen M Vong et al.
Journal of translational medicine, 9, 101-101 (2011-07-05)
Non Obese Diabetic mice lacking B cells (NOD.Igμ(null) mice) do not develop diabetes despite their susceptible background. Upon reconstitution of B cells using a chimera approach, animals start developing diabetes at 20 weeks of age. We have used the spectratyping
Jerzy Dorosz et al.
Scientific reports, 9(1), 4019-4019 (2019-03-13)
The full length human histone 3 lysine 4 demethylase KDM5B (PLU-1/Jarid1B) has been studied using Hydrogen/Deuterium exchange mass spectrometry, homology modelling, sequence analysis, small angle X-ray scattering and electron microscopy. This first structure on an intact multi-domain Jumonji histone demethylase
Xi-Qin Ding et al.
PloS one, 4(10), e7410-e7410 (2009-10-14)
Subretinal delivery of polyethylene glycol-substituted lysine peptide (CK30PEG)-compacted DNA nanoparticles results in efficient gene expression in retinal cells. This work evaluates the ocular safety of compacted DNA nanoparticles. CK30PEG-compacted nanoparticles containing an EGFP expression plasmid were subretinally injected in adult
Radmila Hrdlicková et al.
Molecular and cellular biology, 29(3), 929-941 (2008-12-03)
Telomerase activity is downregulated in somatic cells but is upregulated during the activation of cells of the immune system. The mechanism of this reactivation is not well understood. In this study, we demonstrated that interferon regulatory factor 4 (IRF-4) and
Dayo O Adewole et al.
Science advances, 7(4) (2021-02-02)
For implantable neural interfaces, functional/clinical outcomes are challenged by limitations in specificity and stability of inorganic microelectrodes. A biological intermediary between microelectrical devices and the brain may improve specificity and longevity through (i) natural synaptic integration with deep neural circuitry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service