Skip to Content
Merck
All Photos(2)

Key Documents

A5979

Sigma-Aldrich

Anti-ARP3 antibody, Mouse monoclonal

clone FMS338, purified from hybridoma cell culture

Synonym(s):

ARP3 Detection Antibody, Mouse Anti-ARP3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FMS338, monoclonal

form

buffered aqueous solution

mol wt

antigen 40-45 kDa

species reactivity

human, mouse, rat, canine

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.25-0.5 μg/mL using total cell extract of mouse myoblast C2 cells

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTR3(10096)
mouse ... Actr3(74117)
rat ... Actr3(81732)

General description

Monoclonal Anti-ARP3 (mouse IgG2b isotype) is derived from the hybridoma FMS338 produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with recombinant human ARP3. Arp3 is mapped to human chromosome 2q14.1.

Immunogen

recombinant human ARP3.

Application

Monoclonal Anti-ARP3 antibody produced in mouse has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunocytochemistry
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence
  • immunohistochemistry

Biochem/physiol Actions

The ARP2/3 complex consists of seven distinct proteins (Arp2 and Arp3, p41-Arc, p34-Arc, p21-Arc, p20-Arc, and p16-Arc) that modulate polymerization of cellular actin filaments. ARP3 regulates actin nucleation, and is involved in directing the movement of Listeria monocytogenes in infected host cells. Monoclonal Anti-ARP3 antibody binds to human, canine, rat, and mouse ARP3.
The actin-related protein-2/3 (ARP2/3) is part of Wiskott-Aldrich Syndrome protein (WASP) signaling in mediating polymerization state of actin filaments. It has been found that ARP2/3 complex and its activator spinocerebellar ataxia, autosomal recessive 2 (Scar2) are involved in Golgi polarization in NIH 3T3 cells. However in primary astrocytes, Golgi polarization involves neither actin cytoskeleton or Arp2/3 complex nor any WASP-family of proteins. In a different model, it was found that the ARP2/3 complex was necessary for neutrophil chemotaxis and phagocytosis. By genetic and loss of function studies, it was demonstrated that Scar2 is the major regulator of the ARP2/3 complex rather than the WASP family of proteins, the latter having more restricted roles in specific cellular events.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmental toxicants perturb human Sertoli cell adhesive function via changes in F-actin organization mediated by actin regulatory proteins
Xiao X, et al.
Human Reproduction, 29(6), 1279-1291 (2014)
Genome-wide association study suggested the PTPRD polymorphisms were associated with weight gain effects of atypical antipsychotic medications
Yu H, et al.
Schizophrenia Bulletin, 42(3) (2015)
Identification of the p16-Arc subunit of the Arp 2/3 complex as a substrate of MAPK-activated protein kinase 2 by proteomic analysis
Singh S, et al.
The Journal of biological chemistry, 278(38), 36410-36417 (2003)
Paxillin phosphorylation controls invadopodia/podosomes spatiotemporal organization
Badowski C, et al.
Molecular Biology of the Cell, 19(2), 633-645 (2008)
Kole T Roybal et al.
Science signaling, 9(424), rs3-rs3 (2016-04-21)
Fluorescence microscopy is one of the most important tools in cell biology research because it provides spatial and temporal information to investigate regulatory systems inside cells. This technique can generate data in the form of signal intensities at thousands of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service