Skip to Content
Merck
All Photos(1)

Key Documents

A5855

Sigma-Aldrich

Abz-LFK(Dnp)-OH trifluoroacetate salt

≥97% (HPLC)

Synonym(s):

O-Aminobenzoic acid-Leu-Phe-Lys(DNP)-OH trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H41N7O9 · xC2HF3O2
Molecular Weight:
691.73 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

film

color

yellow

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(=O)c1ccccc1N)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](CCCCNc3ccc(cc3[N+]([O-])=O)[N+]([O-])=O)C(O)=O

InChI

1S/C34H41N7O9.C2HF3O2/c1-21(2)18-28(38-31(42)24-12-6-7-13-25(24)35)32(43)39-29(19-22-10-4-3-5-11-22)33(44)37-27(34(45)46)14-8-9-17-36-26-16-15-23(40(47)48)20-30(26)41(49)50;3-2(4,5)1(6)7/h3-7,10-13,15-16,20-21,27-29,36H,8-9,14,17-19,35H2,1-2H3,(H,37,44)(H,38,42)(H,39,43)(H,45,46);(H,6,7)/t27-,28-,29-;/m0./s1

InChI key

BAXBDLHYKKDHEJ-BJRQXHFHSA-N

Amino Acid Sequence

Abz-Leu-Phe-Lys-DNP

Application

Abz-LFK(Dnp)-OH trifluoroacetate (TFA) salt is an Angiotensin Converting Enzyme (ACE) fluorescent peptide substrate that binds specifically to the C domain of ACE. Abz-LFK(Dnp)-OH TFA has been used to study the development of new antibacterial calixarene derivatives, as well as the efficacy and toxicity of the antimicrobial peptide M33 as a possible antimicrobial agent in lung infections and sepsis.

Biochem/physiol Actions

Abz-LFK(Dnp)-OH is an Angiotensin Converting Enzyme (ACE) fluorescent peptide substrate specific for the C domain for real time fluorescent assay (ACE, Peptidyl Dipeptidase, Kininase II, E.C. 3.4.15.1).

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L-arginine trifluoroacetate salt bridges in its solid state compound: the low-temperature three dimensional structural determination of L-arginine bis(trifluoroacetate) crystal and its vibrational spectral analysis
Sun, Z.H., et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 83, 39-45 (2011)
Guillaume Sautrey et al.
The journal of physical chemistry. B, 115(50), 15002-15012 (2011-11-16)
Tetra-p-guanidinoethylcalix[4]arene trifluoroacetate salt (CX1) was synthesized recently as an antibacterial agent. It showed to be active in vitro against various Gram-positive and Gram-negative bacteria. To get more insight in the mechanism of the biological activity of this derivative, it was
Alessandro Pini et al.
Amino acids, 43(1), 467-473 (2011-10-11)
The tetra-branched peptide M33 (Pini et al. in FASEB J 24:1015-1022, 2010) is under evaluation in animal models for its activity as antimicrobial agent in lung infections and sepsis. The preclinical development of a new drug requires medium-scale manufacture for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service