Skip to Content
Merck

A5602

AMD3100 octahydrochloride hydrate

≥97% (NMR), solid, CXCR4 antagonist

Synonym(s):

1,1′-[1,4-Phenylenebis(methylene)]bis-1,4,8,11-tetraazacyclotetradecane octahydrochloride, JM3100, Plerixafor, SID791

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H54N8 · 8HCl · xH2O
CAS Number:
Molecular Weight:
794.47 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥97% (NMR)
Form:
solid
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AMD3100 octahydrochloride hydrate, ≥97% (NMR), solid

Quality Segment

assay

≥97% (NMR)

form

solid

storage condition

desiccated

solubility

H2O: ≥10 mg/mL, clear

originator

Sanofi Aventis

storage temp.

−20°C

SMILES string

Cl[H].Cl[H].Cl[H].Cl[H].Cl[H].Cl[H].Cl[H].Cl[H].[H]O[H].C1CNCCNCCCN(CCNC1)Cc2ccc(CN3CCCNCCNCCCNCC3)cc2

InChI

1S/C28H54N8.8ClH.H2O/c1-9-29-15-17-31-13-3-21-35(23-19-33-11-1)25-27-5-7-28(8-6-27)26-36-22-4-14-32-18-16-30-10-2-12-34-20-24-36;;;;;;;;;/h5-8,29-34H,1-4,9-26H2;8*1H;1H2

InChI key

GKFHDCZYJHUPEN-UHFFFAOYSA-N

Application

AMD3100 is a highly specific chemokine receptor CXCR4 antagonist. AMD3100 has been used in a study to determine the blockade of CXCR4 in oral squamous cell carcinoma and inhibition of lymph node metastases.

Biochem/physiol Actions

AMD3100 is a highly specific chemokine receptor CXCR4 antagonist.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library