A5596
Greiner multiwell plate sealers
EASYseal™ sealing film, transparent, standard adhesive foil on carrier paper
Synonym(s):
multiwell plate sealing films, sealing films, sealing tape
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.01
Recommended Products
material
PET
polyester film
sterility
non-sterile
packaging
pack of 100
manufacturer/tradename
Greiner 676001
W × L
7.938 cm × 13.1 cm
color
clear
suitability
suitable for (for coverage/ storage of microplates)
Looking for similar products? Visit Product Comparison Guide
General description
Greiner microplate sealers are RNase free, DNase free and nonpyrogenic. They feature a temperature tolerance range of -70 °C up to 110 °C.
Standard adhesive-backed microplate sealer suitable for all traditional laboratory purposes where protection of sample integrity and evaporation prevention are desired.
Material:
Foil: Polyethyleneterephtalat (PET)
Glue: Acrylate Solvent
Paper: Silicone-paper KV 75
Material:
Foil: Polyethyleneterephtalat (PET)
Glue: Acrylate Solvent
Paper: Silicone-paper KV 75
Application
Greiner multiwell plate sealers have been used for isolating the ends of mounted samples to prevent the conductive nature of metal fixtures. The study was done to compare the electrical properties between the bulk conductive-PDMS (polydimethylsiloxane) material and the microengineered electrodes under stretch. Greiner multiwell plate sealers have also been used for the sealing of 96-well V-bottom polypropylene plates for high-throughput analysis of the plasma N-glycome via UHPLC (ultrahigh performance liquid chromatography).
Legal Information
EASYseal is a trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Microengineered Conductive Elastomeric Electrodes for Long-Term Electrophysiological Measurements with Consistent Impedance under Stretch.
Hu D, et al.
Sensors (Basel, Switzerland), 15, 26906-26920 (2015)
High-Throughput Analysis of the Plasma N-Glycome by UHPLC.
Adamczyk B, et al.
Methods in Molecular Biology, 1503, 97-108 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service