Skip to Content
Merck
All Photos(3)

Key Documents

A4174

Sigma-Aldrich

Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Goat IgG (whole molecule)–HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

goat

should not react with

human

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibodies regulate several functions such as complement activation and phagocytosis. Thus they play a crucial role in facilitating cytological immune responses. Anti-goat IgG (whole molecule)–peroxidase antibody can be used as a secondary antibody in ovarian tissue microarray. Rabbit anti-goat IgG (whole molecule)-peroxidase antibody reacts specifically with all goat IgG but shows no reactivity with human serum proteins.

Immunogen

Purified goat IgG.

Application

Anti-Goat IgG (whole molecule)-Peroxidase antibody produced in rabbit is suitable for use in immunoblot and immunohistochemistry. The product can also be used for direct ELISA (1:10,000).
The presence of pleiotrophin in HUVEC cell culture medium was analyzed by western blot using HRP-conjugated rabbit anti-goat IgGas the secondary at a dilution of 1:7500 in TBST.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% MIT.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebekka Mauser et al.
Epigenetics & chromatin, 10(1), 45-45 (2017-09-28)
Histone post-translational modifications (PTMs) play central roles in chromatin-templated processes. Combinations of two or more histone PTMs form unique interfaces for readout and recruitment of chromatin interacting complexes, but the genome-wide mapping of coexisting histone PTMs remains an experimentally difficult task.
Peter Horak et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 11(24 Pt 1), 8585-8591 (2005-12-20)
Epithelial ovarian cancer is the most common cause of mortality from gynecologic malignancies. Due to advanced stage at diagnosis, most patients need systemic treatment in addition to surgery. Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) is a member of the
Sanja Cabric et al.
Tissue engineering. Part A, 16(3), 961-970 (2009-12-22)
In pancreatic islet transplantation, early revascularization is necessary for long-term graft function. We have shown in in vitro and in vivo models that modification with surface-attached heparin protects the islets from acute attack by the innate immune system of the
Evangelia Poimenidi et al.
Anticancer research, 29(1), 349-354 (2009-04-01)
Despite the fact that pleiotrophin (PTN) exhibits important biological activities related to tumor growth and angiogenesis, little is known about the regulation of its expression. In the present work, the effect of serum on PTN expression and secretion in the
Leanne M Mullen et al.
Journal of materials science. Materials in medicine, 26(1), 5325-5325 (2015-01-13)
Tissue engineering is a promising technique for cartilage repair. Toward this goal, a porous collagen-glycosaminoglycan (CG) scaffold was loaded with different concentrations of insulin-like growth factor-1 (IGF-1) and evaluated as a growth factor delivery device. The biological response was assessed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service