Skip to Content
Merck
All Photos(1)

Key Documents

A3356

Sigma-Aldrich

Anti-Beta (β)-Amyloid antibody

rabbit polyclonal

Synonym(s):

Anti-A4 Amyloid protein, Anti-ABETA, Anti-ABPP, Anti-AD1, Anti-APPI, Anti-Amyloid β A4 protein, Anti-Amyloid-β protein (ABP)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-β-Amyloid (22-35) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~4 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

technique(s)

indirect ELISA: 0.2-0.4 μg/mL using β-amyloid peptide (22-35)
western blot: 0.25-0.5 μg/mL using β-amyloid peptide (1-40)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APP(351)
mouse ... App(11820)
rat ... App(54226)

General description

Rabbit anti-β-amyloid (22-35) antibody specifically recognizes β-amyloid (22-35), β-amyloid (25-35), β-amyloid (1-40) and β-amyloid (1-42) and does not react with β-amyloid (32-35) and β-amyloid (35-25) by ELISA. The antibody detects β-amyloid (1-40) by immunoblotting (approx. 4kDa). Staining of the β-amyloid (1-40) band in immunoblotting is specifically inhibited by the immunizing peptide.
Cleavage of the amyloid precursor protein (APP) by γ-secretase produces the small fragment amyloid β (Aβ) peptide. Cleavage of APP occurs mostly at residue 40 but to a lesser extent residue 42. The amyloid β peptide contains a ~100 kDa soluble N-terminal fragment, and intracellular C-terminal fragments (CTFs) bearing the complete Aβ domain.
Proteolytic cleavage of the amyloid precursor protein (APP) produces the small fragment amyloid β peptide. Cleavage of APP occurs mostly at residue 40 but to a lesser extent residue 42. Aggregates of amyloid β peptide are found deposited in the brains of Alzheimer′s patients.

Immunogen

synthetic peptide corresponding to amino acids 22-35 of human β-amyloid (1-40) fragment, conjugated to KLH. This sequence corresponds to amino acids 693-706 of the human amyloid precursor protein APP, and is identical in mouse and rat APP.

Application

Rabbit anti-β-amyloid (22-35) antibody can be used for ELISA at a concentration of 0.2-0.4μg/mL using β-amyloid peptide (22-35). The product can also be used for western blot at 0.25-0.5μg/mL using β-amyloid peptide (1-40).
Anti-β-Amyloid (22-35) antibody produced in rabbit has been used in:
  • western blot
  • dot-blot assay
  • indirect enzyme linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Secreted Aβ lead to synaptic and neuritic compromise and glial activation. Aggregates of amyloid β peptide are found deposited in the brains of Alzheimer′s patients. Aβ(25-35) and Aβ(22-35) fragments are highly toxic segments of β-amyloid peptides that promote inflammatory processes in astrocytes and fibrillary aggregation of Aβ, thus representing a promising therapeutic target for Alzheimer′s.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide and 1% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms of transthyretin inhibition of $\beta$-amyloid aggregation in vitro
Li X, et al.
The Journal of Neuroscience, 33(50), 19423-19433 (2013)
IL-1beta, TNF-alpha and Sambucus nigra Reactive Serum Proteins as Biomarkers of Mild Cognitive Impairment and Alzheimer Disease Progression
Castillo LM< et al.
Advances in Alzheimer's disease, 4(04), 99-99 (2015)
An Overview of APP Processing Enzymes and Products
Chow VW, et al.
Neuromolecular Medicine, 12, 1-1 (2010)
Xinyi Li et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(50), 19423-19433 (2013-12-18)
Tissue-specific overexpression of the human systemic amyloid precursor transthyretin (TTR) ameliorates Alzheimer's disease (AD) phenotypes in APP23 mice. TTR-β-amyloid (Aβ) complexes have been isolated from APP23 and some human AD brains. We now show that substoichiometric concentrations of TTR tetramers
Mechanisms of Amyloid-beta Peptide Clearance: Potential Therapeutic Targets for Alzheimer's Disease
Yoon SS, et al.
Biomolecules & Therapeutics, 20, 245-245 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service