Skip to Content
Merck
All Photos(1)

Key Documents

A2836

Sigma-Aldrich

Ansamitocin P-3 from Actinosynnema pretiosum

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H43ClN2O9
CAS Number:
Molecular Weight:
635.14
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥90% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

CO[C@@H]1\C=C\C=C(C)\Cc2cc(OC)c(Cl)c(c2)N(C)C(=O)C[C@H](OC(=O)C(C)C)[C@]3(C)O[C@@H]3[C@H](C)[C@@H]4C[C@@]1(O)NC(=O)O4

InChI

1S/C32H43ClN2O9/c1-17(2)29(37)43-25-15-26(36)35(6)21-13-20(14-22(40-7)27(21)33)12-18(3)10-9-11-24(41-8)32(39)16-23(42-30(38)34-32)19(4)28-31(25,5)44-28/h9-11,13-14,17,19,23-25,28,39H,12,15-16H2,1-8H3,(H,34,38)/b11-9+,18-10+/t19-,23+,24-,25+,28-,31+,32+/m1/s1

InChI key

OPQNCARIZFLNLF-KUDLRRJMSA-N

Biochem/physiol Actions

Fungal metabolite with antineoplastic, antimitotic activity. Binds to tubulin and inhibits vinblastine-induced spiral formation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel L Segraves et al.
Magnetic resonance in chemistry : MRC, 50(3), 256-259 (2012-03-01)
During impurity analysis of maytansinol (2), produced from the reduction of ansamitocin P-3 (AP-3, 1), a surprisingly stable acyclic hemiacetal (4) was isolated. A combination of 1D and 2D NMR experiments, along with liquid chromatography-mass spectrometry data was used to
Tobias Knobloch et al.
Chembiochem : a European journal of chemical biology, 12(4), 540-547 (2012-01-13)
New ansamitocin derivatives were prepared by feeding aminobenzoic acid derivatives to cultures of Actinosynnema pretiosum HGF073, a mutant strain blocked in the biosynthesis of the required 3-amino-5-hydroxybenzoic acid (AHBA) starter unit. Use of several aminobenzoic acids as precursors led to
Florian Taft et al.
Journal of the American Chemical Society, 131(11), 3812-3813 (2009-03-19)
The timing of introduction of the unusually placed Delta(11,13) diene system in ansamitocin (AP) biosynthesis was probed by synthesizing optically active potential tri- and tetraketide intermediates as their SNAC thioesters. An AP-nonproducing mutant Actinosynnema pretiosum was complemented by the R
Peiji Zhao et al.
Chemistry & biology, 15(8), 863-874 (2008-08-30)
Ansamitocins are potent antitumor maytansinoids produced by Actinosynnema pretiosum. Their biosynthesis involves the initial assembly of a macrolactam polyketide, followed by a series of postpolyketide synthase (PKS) modifications. Three ansamitocin glycosides were isolated from A. pretiosum and fully characterized structurally
Juan Ma et al.
Archives of pharmacal research, 30(6), 670-673 (2007-08-08)
By using preparative TLC as the critical isolation procedure, two compounds, including one new amide N-glycosides of ansamitocin (2), were isolated from Actinosynnema pretiosum. The compounds were elucidated as N-demethyl-N-beta-D-glucopyranosyl ansamitocin P-2, named ansamitocinoside P-2 (1), and N-demethyl-N-beta-D-glucopyranosyl ansamitocin P-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service