A2636
S-(2-Aminoethyl)-L-cysteine hydrochloride
≥98% (TLC)
Synonym(s):
L-4-Thialysine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H12N2O2S · HCl
CAS Number:
Molecular Weight:
200.69
Beilstein:
3697262
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
S-(2-Aminoethyl)-L-cysteine hydrochloride, ≥98% (TLC)
Assay
≥98% (TLC)
form
powder
color
white to off-white
storage temp.
2-8°C
SMILES string
Cl.NCCSC[C@H](N)C(O)=O
InChI
1S/C5H12N2O2S.ClH/c6-1-2-10-3-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1
InChI key
CVHKULVNPGAEQM-WCCKRBBISA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
S-(2-Aminoethyl)-L-cysteine (AEC) hydrochloride is used as a lysine analogue for comparative analysis with other lysine analogues. S-(2-Aminoethyl)-L-cysteine is an alternative substrate useful for characterizing lysine cyclodeaminase from Streptomyces pristinaespiralis. AEC may be used as a non-antibiotic selection agent for genetically engineered soybeans expressing a lysine insensitive DHPS gene. AEC is being studied as an amino acid antibiotic.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Asri Peni Wulandari et al.
FEBS letters, 522(1-3), 35-40 (2002-07-04)
In Thermus thermophilus homocitrate synthase (HCS) catalyzes the initial reaction of lysine biosynthesis through alpha-aminoadipic acid, synthesis of homocitrate from 2-oxoglutarate and acetyl-CoA. HCS is strongly inhibited by lysine, indicating that the biosynthesis is regulated by the endproduct at the
M N Cahyanto et al.
Journal of applied microbiology, 102(3), 674-679 (2007-02-21)
To enhance L-lysine secretion in Lactobacillus plantarum. An S-2-aminoethyl-L-cystein (AEC)-resistant mutant of L. plantarum was isolated, and it produced L-lysine at considerably higher level than the parent strain. Aspartokinase in the mutant has been desensitized to feedback inhibition by L-lysine.
Dasantila Golemi-Kotra et al.
The Journal of biological chemistry, 279(33), 34665-34673 (2004-05-21)
Beta-lactamases and penicillin-binding proteins are bacterial enzymes involved in antibiotic resistance to beta-lactam antibiotics and biosynthetic assembly of cell wall, respectively. Members of these large families of enzymes all experience acylation by their respective substrates at an active site serine
Do Youn Jun et al.
Biochemical pharmacology, 66(12), 2291-2300 (2003-11-26)
We first report the mechanism for the inhibitory effect of the lysine analog, thialysine on human acute leukemia Jurkat T cells. When Jurkat T cells were treated with thialysine (0.32-2.5 mM), apoptotic cell death along with several biochemical events such
Ulf Göransson et al.
Analytical biochemistry, 318(1), 107-117 (2003-06-05)
The expression of cyclotides-macrocyclic plant peptides-was profiled in six violets, Viola cotyledon, V. biflora, V. arvensis, V. tricolor, V. riviniana, and V. odorata, by LC-MS. All were found to express notably complex mixtures, with single species containing >50 cyclotides. To
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service