Skip to Content
Merck
All Photos(1)

Key Documents

A2404

Sigma-Aldrich

Alcohol Oxidase solution from Pichia pastoris

buffered aqueous solution, 10-40 units/mg protein (biuret)

Synonym(s):

Alcohol:oxygen oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

buffered aqueous solution

specific activity

10-40 units/mg protein (biuret)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Alcohol oxidase (AOX) is a homo-octamer, which has a molecular weight of 600-kDa. It has eight flavin adenine dinucleotide (FAD) cofactors. It belongs to the glucose-methanol-choline (GMC) oxidoreductase family.

Application

Alcohol Oxidase may be used to study protein translocation into peroxisomes. Product A2404 is from Pichia pastoris. It has been used for the bacterial expression and immunological verification of Hv-p68 cDNA clones .
Alcohol Oxidase solution from Pichia pastoris has been used in cotton pectin methylesterase (PME) enzyme assay. It has also been used to immobilize onto polyaniline (PANI) film as a visual biosensor in dip-stick format for the determination of ethanol in fermented beverage samples.

Biochem/physiol Actions

Alcohol oxidase catalyzes the oxidation of short-chain, primary, aliphatic alcohols to their respective aldehydes. It has the highest affinity for methanol. Alcohol oxidase is involved in the metabolism of methanol, which is important for yeasts that utilize methanol as their sole energy source. Alcohol oxidase is primarily found in the peroxisome but may be present in the cytoplasm as well.

Unit Definition

One unit will oxidize 1.0 μmole of methanol to formaldehyde per min at pH 7.5 at 25 °C.

Physical form

Solution in 30% sucrose with 0.1 M phosphate buffer at pH 8.0

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuan Wang et al.
Journal of experimental botany, 71(9), 2629-2640 (2020-02-02)
De-esterification of homogalacturonan (HG) is thought to stiffen pectin gels and primary cell walls by increasing calcium cross-linking between HG chains. Contrary to this idea, recent studies found that HG de-esterification correlated with reduced stiffness of living tissues, measured by
Pectin methylesterase and pectin remodelling differ in the fibre walls of two gossypium species with very different fibre properties
Liu Q, et al.
PLoS ONE, 8(6), e65131-e65131 (2013)
Structure of alcohol oxidase from Pichia pastoris by cryo-electron microscopy
Vonck J, et al.
PLoS ONE, 11(7), e0159476-e0159476 (2016)
A I Soldevila et al.
The Journal of biological chemistry, 276(7), 4652-4661 (2000-11-01)
We have cloned and sequenced a novel alcohol oxidase (Hv-p68) from the filamentous fungus Helminthosporium (Cochliobolus) victoriae that copurifies with mycoviral double-stranded RNAs. Sequence analysis revealed that Hv-p68 belongs to the large family of FAD-dependent glucose methanol choline oxidoreductases and
A simple visual ethanol biosensor based on alcohol oxidase immobilized onto polyaniline film for halal verification of fermented beverage samples
Kuswandi B, et al.
Sensors, 14(2), 2135-2149 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service