Skip to Content
Merck
All Photos(1)

Key Documents

A1602

Sigma-Aldrich

Amyloglucosidase from Aspergillus niger

ammonium sulfate suspension, ≥40 units/mg protein

Synonym(s):

1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

ammonium sulfate suspension

Quality Level

specific activity

≥40 units/mg protein

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Amyloglucosidase, a disaccharide enzyme is a member of the alpha-glucosidase type. It is produced by several species of Aspergillus genus.

Application

Amyloglucosidase from Aspergillus niger is used to hydrolyze α-D-glucosides. It may be used in the brewing of beer and in the production of bread and juices. Amyloglucosidase, from Sigma, has been used to hydrolyze glycogen into glucose monomers in order to study lipid accumulation in skeletal muscle .
Amyloglucosidase from Aspergillus niger has been used to liberate free glucose in the Belgica antarctica larva homogenate. It has also been used in digestive enzyme mixture for starch digestion.

Biochem/physiol Actions

Amyloglucosidase from Aspergillus niger is capable of hydrolyzing the α-D-(1-4), the α-D-(1-6), and the α-D-(1-3) glucosidic bonds of oligosaccharides .

Unit Definition

One unit will liberate 1.0 mg of glucose from starch in 3 min at pH 4.5 at 55 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4 solution, pH approx. 6.0

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method for routine measurements of total sugar and starch content in woody plant tissues
Chow PS and Landhausser SM
Tree physiology, 24(10), 1129-1136 (2004)
Biodiversity: New Leads for the Pharmaceutical and Agrochemical Industries, 183-183 (2000)
Survival and energetic costs of repeated cold exposure in the Antarctic midge, Belgica antarctica: a comparison between frozen and supercooled larvae
Teets NM, et al.
The Journal of Experimental Biology, 214(5), 806-814 (2011)
Christina Pedersen et al.
PloS one, 13(3), e0194468-e0194468 (2018-03-21)
Insulin is a vital part of diabetes treatment, whereas glucagon is primarily used to treat insulin-induced hypoglycemia. However, glucagon is suggested to have a central role in the regulation of body weight, which would be beneficial for diabetic patients. Since
Jane Usher et al.
Applied and environmental microbiology, 75(13), 4573-4579 (2009-05-12)
Yeasts used in the production of lagers contain complex allopolyploid genomes, resulting from the fusion of two different yeast species closely related to Saccharomyces cerevisiae and Saccharomyces bayanus. Recombination between the homoeologous chromosomes has generated a number of hybrid chromosomes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service