Skip to Content
Merck
All Photos(1)

Key Documents

A129

Sigma-Aldrich

Amoxapine

Synonym(s):

2-Chloro-11-(1-piperazinyl)dibenz[b,f][1,4]oxazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16ClN3O
CAS Number:
Molecular Weight:
313.78
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

solubility

methanol: soluble

originator

Wyeth

SMILES string

Clc1ccc2Oc3ccccc3N=C(N4CCNCC4)c2c1

InChI

1S/C17H16ClN3O/c18-12-5-6-15-13(11-12)17(21-9-7-19-8-10-21)20-14-3-1-2-4-16(14)22-15/h1-6,11,19H,7-10H2

InChI key

QWGDMFLQWFTERH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amoxapine has been used as an antidepressant drug to test its effect on locomotion and egg release in response to gain-of-functional mutations in potassium (K+) channels (unc-58) of C. elegans. It has also been used as an antipsychotic drug to test its effect on the viability of glioblastoma cells.

Biochem/physiol Actions

Amoxapine, a structural analog of clozapine, is a human ether a-go-go (hERG) channel blocker. It is also an N-methylated metabolite of loxapine. It is a tricyclic antidepressant that inhibits the uptake of norepinephrine and blocks 5- hydroxytryptamine (HT2 ) serotonergic receptors.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K S Merigian et al.
Academic emergency medicine : official journal of the Society for Academic Emergency Medicine, 2(2), 128-133 (1995-02-01)
Tonic-clonic seizure activity is a recognized complication of amoxapine overdose. Refractory drug-induced status epilepticus is associated with significant morbidity and mortality. Standard regimens for controlling status epilepticus may be ineffective for aborting drug-induced seizures. The authors report the case of
Manju Gupta et al.
Journal of separation science, 33(23-24), 3774-3780 (2010-11-18)
A new, selective and sensitive method has been developed for the determination of tricyclic antidepressant drugs, amoxapine and nortriptyline, in human blood plasma and serum, involving their reaction with allyl isothiocyanate and extraction of thiourea derivatives with water-miscible organic solvent
Hiroaki Kobayashi et al.
Journal of pharmaceutical and biomedical analysis, 54(1), 258-263 (2010-09-15)
Phase-solubility studies have been used to evaluate the solubilizing effects of cyclodextrins (CDs) on lipophilic, water-insoluble drugs. However, large amounts of CDs and drugs are required to measure solubility by phase-solubility studies. Thus, more efficient approaches to evaluate the interaction
J E Dugas et al.
Drug intelligence & clinical pharmacy, 16(3), 199-204 (1982-03-01)
Amoxapine is a tricyclic antidepressant agent, which is chemically related to the antipsychotic agent loxapine, but which appears to block selectively the neuronal reuptake of norepinephrine; it is qualitatively similar to desipramine. In studies of patients with mixed depressive illnesses
T A Ban et al.
International pharmacopsychiatry, 15(3), 166-170 (1980-01-01)
Amoxapine, a new antidepressant, is a tricyclic debenzoxazepine compound, the demethylated metabolite of the neuroleptic loxapine. In animal pharmacological studies, amoxapine has shown striking similarities to imipramine. In contrast to the prototype antidepressant drug, however, amoxapine does not interact with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service