Skip to Content
Merck
All Photos(1)

Documents

A1131

Sigma-Aldrich

L-Anserine nitrate salt

≥98%

Synonym(s):

β-Ala-3-methyl-His nitrate salt, β-Alanyl-3-methyl-L-histidine nitrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N4O3 · HNO3
CAS Number:
Molecular Weight:
303.27
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Anserine nitrate salt, hydroxyl radical scavenger

Quality Level

Assay

≥98%

form

powder

color

white

application(s)

detection

SMILES string

O[N+]([O-])=O.Cn1cncc1C[C@H](NC(=O)CCN)C(O)=O

InChI

1S/C10H16N4O3.HNO3/c1-14-6-12-5-7(14)4-8(10(16)17)13-9(15)2-3-11;2-1(3)4/h5-6,8H,2-4,11H2,1H3,(H,13,15)(H,16,17);(H,2,3,4)/t8-;/m0./s1

InChI key

OLWOKAYJAHHSNY-QRPNPIFTSA-N

Biochem/physiol Actions

L-Anserine s a dipeptide found in most animal tissues. In model systems it is a potent antioxidant and scavener of hydroxyl radicals. It inhibits nonenzymatic protein glycation induced by aldose and ketose reducing sugars. It is much less potent than L-carnosine as an inhibitor of protein-protein crosslinking.

Linkage

Anserine is a naturally occurring methylated analog of carnosine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wieslaw Kopec et al.
Antioxidants (Basel, Switzerland), 9(11) (2020-11-12)
The objective of the study was to test the effect of diets supplemented with β-alanine, L-histidine, and carnosine on the histidine dipeptide content and the antioxidative status of chicken breast muscles and blood. One-day-old Hubbard Flex male chickens were assigned
Verena Peters et al.
Amino acids, 42(6), 2411-2416 (2011-08-13)
Recently, we identified an allelic variant of human carnosinase 1 (CN1) that results in increased enzyme activity and is associated with susceptibility for diabetic nephropathy in humans. Investigations in diabetic (db/db) mice showed that carnosine ameliorates glucose metabolism effectively. We
Angelo Zinellu et al.
Talanta, 84(3), 931-935 (2011-04-13)
A field amplified sample injection (FASI) capillary electrophoresis method with UV detection was developed for the separation and detection of carnosine-related peptides carnosine (Car), anserine (Ans) and homocarnosine (Hcar). The imidazole dipeptides were baseline-separated within 10 min by using 50
Daiki Kubomura et al.
Nutritional neuroscience, 13(4), 183-188 (2010-07-31)
Anserine and L-carnosine are similar dipeptides synthesized by muscles of vertebrates. The functional role of anserine is unknown, although previous studies showed hypoglycemic effects of carnosine through autonomic nerves. Thus, we evaluated the effects of anserine on blood glucose levels
Effect of anserine ingestion on the hyperglycemia and autonomic nerves in rats and humans.
Kubomura D, Matahira Y, Nagai K, Niijima A.
Nutritional Neuroscience, 13, 1230128-1230128 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service