Skip to Content
Merck

A0972

Apolipoprotein A-II from human plasma

≥95% (SDS-PAGE), buffered aqueous solution

Synonym(s):

Apo A-II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human plasma

Quality Segment

assay

≥95% (SDS-PAGE)

form

buffered aqueous solution

mol wt

17.38 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... APOA2(336)

General description

Forms dimers in aqueous solutions with an association constant of 5 × 104 M-1. The association is accompanied by an enhancement of ~35% in α-helical content.

Application

Apo A-II constitutes ~20% of the protein moiety of high density lipoprotein (HDL).

Biochem/physiol Actions

Unlike Apo A-I, Apo A-II possesses no activating properties for lectin-cholesterol acyltransferase (LCAT). Furthermore, it was found that Apo A-II inhibits the activation of LCAT by Apo A-I.

Physical form

Solution in 10 mM ammonium bicarbonate.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library