Skip to Content
Merck
All Photos(1)

Key Documents

A0731

Sigma-Aldrich

Anti-ATG5 (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Atg5 (C-terminal) antibody produced in rabbit, Anti-ASP, Anti-Apg5, Anti-Apg5L, Anti-Apoptosis-specific protein, Anti-Autophagy protein 5-like

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen ~56 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: 1-2 μg/mL using whole extract of mouse liver or human U87 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATG5(9474)
mouse ... Atg5(11793)

General description

ATG5 (autophagy related 5) is located on human chromosome band 6q21.
ATG5 was found localized to double FYVE domain containing protein 1 (DFCP1)- specific omegasomes. [1]
Macroautophagy, usually referred to as autophagy, is a major pathway for bulk degradation of cytoplasmic constituents and organelles. Autophagy is an evolutionary conserved pathway seen in all eukaryotic cells. At least 16 ATG genes, required for autophagosome formation were identified in yeast by genetic screens. For many of these genes, related homologs have been identified in mammals. Atg5, also known as Apg5, is a 32 kDa protein essential for autophagy. Atg5 is covalently modified by Atg12, a ubiquitin-like modifier. This conjugation reaction requires ATP and two enzymes, Atg7 and Atg10, which are E1- and E2-like enzymes, respectively. The Atg12-Atg5 conjugate interacts non-covalently with Atg16. The Atg12-Atg5-Atg16 complex localizes to autophagosome precursors and plays an essential role in autophagosome formation. It has been reported that in addition to the role of Atg5 in the formation of autophagosomes, an Atg5 fragment produced by calpain cleavage has pro-apoptotic properties.
Rabbit polyclonal anti-ATG5 (C-terminal) antibody recognizes mouse and human Atg5-Atg12 complex by immunoblotting, ~56 kDa. Detection of the Atg5-Atg12 complex band by immunoblotting is specifically inhibited with the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 262-275 of mouse Atg5, conjugated to KLH via an N-terminal cysteine residue. The corresponding sequence differs by one amino acid in human.

Application

Anti-ATG5 (C-terminal) antibody has been used in immunohistochemical (IHC) staining.
Rabbit anti-ATG5 (C-terminal) antibody has been used for western blot analysis in U2OS cells. The antibody has also been used for immunoblot assays in CT26 and MCA205 cells.
Rabbit polyclonal anti-ATG5 (C-terminal) antibody is used to tag Atg5-Atg12 complex for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of Atg5-Atg12 complex in the formation and function of autophagosomes. Rabbit anti-ATG5 was used as primary antibody for immunocytochemistry on HEK293 and Cos7 cells. Cells were fixed in 3.7% formaldehyde in 200 mM Hepes prior to antibody treatment.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth L Axe et al.
The Journal of cell biology, 182(4), 685-701 (2008-08-30)
Autophagy is the engulfment of cytosol and organelles by double-membrane vesicles termed autophagosomes. Autophagosome formation is known to require phosphatidylinositol 3-phosphate (PI(3)P) and occurs near the endoplasmic reticulum (ER), but the exact mechanisms are unknown. We show that double FYVE
Ashkan Mortezavi et al.
Oncotarget, 8(19), 31765-31774 (2017-04-21)
Autophagy is a catabolic cellular process used for degradation of cytoplasmic organelles and preservation of cell viability. In this study we aimed to analyse the level of autophagy markers in benign and malignant prostate tissue and to evaluate the prognostic
Metastatic risk and resistance to BRAF inhibitors in melanoma defined by selective allelic loss of ATG5
Garcia F M, et al.
Autophagy, 12(10), 1776-1790 (2016)
S Polager et al.
Oncogene, 27(35), 4860-4864 (2008-04-15)
The retinoblastoma pathway is often inactivated in human tumors resulting in deregulated E2F activity that can induce both proliferation and cell death. Although the role of E2F in apoptosis is well characterized, little is known regarding its putative participation in
Ying Gao et al.
Cell death & disease, 7(8), e2330-e2330 (2016-08-12)
Autophagy is an evolutionarily conserved catabolic process by which cells degrade intracellular proteins and organelles in the lysosomes. Canonical autophagy requires all autophagy proteins (ATGs), whereas noncanonical autophagy is activated by diverse agents in which some of the essential autophagy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service