Skip to Content
Merck
All Photos(1)

Key Documents

A0606

Sigma-Aldrich

Aurothioglucose hydrate

≥96% (titration)

Synonym(s):

Gold thioglucose, Solganal, Solganol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11AuO5S · xH2O
CAS Number:
Molecular Weight:
392.18 (anhydrous basis)
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥96% (titration)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

O.OC[C@H]1O[C@H](S[Au])[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O5S.Au.H2O/c7-1-2-3(8)4(9)5(10)6(12)11-2;;/h2-10,12H,1H2;;1H2/q;+1;/p-1/t2-,3-,4+,5-,6-;;/m1../s1

InChI key

SGVIEHTYEMEDRR-PKXGBZFFSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Aurothioglucose hydrate has been used in redox stress survival assay in human cell lines and for inducing obese phenotype in mice.

Biochem/physiol Actions

Aurothioglucose, a gold compound used clinically to treat rheumatoid arthritis, has recently been found to be a potent PKCiota-Par6 interaction inhibitor, with an IC50 approximately 1 μM. Disruption of this interaction disrupts a rac1 signaling pathway that is required for transformed growth in non-small-cell lung cancer.

Features and Benefits

This compound is featured on the PKC page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric N van Roon et al.
The Journal of rheumatology, 32(6), 1026-1030 (2005-06-09)
For reasons of insufficient quality of the raw material, aurothioglucose was withdrawn from the Dutch market at the end of 2001. Aurothiomalate became available as an alternative preparation. We followed a cohort of patients during the first year after switching
Kouichi Yoshinari et al.
Pharmaceutical research, 23(6), 1188-1200 (2006-05-23)
Changes in physiological, pathophysiological, and/or nutritional conditions often alter the expression of drug-metabolizing enzymes. In this study, we investigated obesity-induced changes in hepatic cytochrome P450 (P450) levels using nutritionally obese mice. To induce obesity, mice were fed a high-fat diet
Mitsunari Ogasawara et al.
Liver international : official journal of the International Association for the Study of the Liver, 31(4), 542-551 (2011-03-09)
The search for effective treatments of non-alcoholic steatohepatitis (NASH), now the most common chronic liver disease in affluent countries, is hindered by a lack of animal models having the range of anthropometric and pathophysiological features as human NASH. To examine
P Huypens et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 38(8), 486-490 (2006-08-31)
Following the chemically-induced lesion of the ventromedial nucleus, gold-thioglucose treated rodents display hypothalamic leptin resistance, hyperphagia, hyperinsulinemia and obesity. Despite the exuberant hyperinsulinemia following gold-thioglucose treatment, systemic insulin sensitivity is preserved during the early phase of the obesity syndrome, resulting
Hiroshi Karasawa et al.
Obesity (Silver Spring, Md.), 19(3), 514-521 (2010-08-14)
An obesity-induced diabetes model using genetically normal mouse strains would be invaluable but remains to be established. One reason is that several normal mouse strains are resistant to high-fat diet-induced obesity. In the present study, we show the effectiveness of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service