Skip to Content
Merck
All Photos(1)

Documents

86C

Sigma-Aldrich

Leukocyte Alkaline Phosphatase Kit

based on naphthol AS-BI and fast blue BB salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
eCl@ss:
42010105
NACRES:
NA.47

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Leukocyte Alkaline Phosphatase (LAP) kits are intended for the qualitative demonstration of alkaline phosphatase activity in white blood cells.
Peripheral blood or bone marrow preparations are fixed to a microscope slide. The film is then incubated in a mixture of naphthol AS-BI alkaline solution with fast blue BB. The resulting insoluble diffuse, blue dye deposit indicates sites of alkaline phosphatase activity.

Kit Components Only

Product No.
Description

  • Citrate Solution (915) 50 mL

  • FBB-Alkaline Solution (863) 10 mL

  • Naphthol AS-BI Alkaline Solution (861) 10 mL

  • Neutral Red Solution (N6264) 50 mL

  • Sodium Nitrite Solution (914) 10 mL

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory M Cooper et al.
Tissue engineering. Part A, 16(5), 1749-1759 (2009-12-24)
The purpose of this study was to demonstrate spatial control of osteoblast differentiation in vitro and bone formation in vivo using inkjet bioprinting technology and to create three-dimensional persistent bio-ink patterns of bone morphogenetic protein-2 (BMP-2) and its modifiers immobilized
Ming-Tzu Tsai et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 27(9), 1169-1174 (2009-03-11)
Human mesenchymal stem cells (hMSCs) are a promising candidate cell type for regenerative medicine and tissue engineering applications by virtue of their capacity for self-renewal and multipotent differentiation. Our intent was to characterize the effect of pulsed electromagnetic fields (PEMFs)
Ankur Sharma et al.
Cytotechnology, 72(3), 489-497 (2020-03-04)
In the present study, we used a serum-free culture media to propagate goat putative spermatogonial stem cells (SSCs) and evaluated the effect of crucial growth factors on relative expression of some SSC markers and self-renewal related genes. The enriched SSCs
Ariana Darcy et al.
Bone, 50(6), 1294-1303 (2012-03-17)
Bone homeostasis can be compromised by an increase in osteoclast-mediated resorption and/or a decrease in osteoblast-mediated bone deposition. While many efforts have focused on treating osteoclast resorption, there has been less emphasis on identifying strategies for promoting osteoblast function. Herein
Barbara Klotz et al.
PloS one, 7(1), e29959-e29959 (2012-01-14)
1,25-dihydroxyvitamin D3 (1,25D3) was reported to induce premature organismal aging in fibroblast growth factor-23 (Fgf23) and klotho deficient mice, which is of main interest as 1,25D3 supplementation of its precursor cholecalciferol is used in basic osteoporosis treatment. We wanted to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service