Skip to Content
Merck
All Photos(1)

Key Documents

86453

Sigma-Aldrich

TERGITOL TMN 6

Liquid, 800 ppm, cloud point 36 °C

Synonym(s):

Trimethylnonylpolyethylene glycol, TERGITOL TMN-6 (90% aqueous) surfactant, Polyethylene glycol trimethylnonyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25

description

non-ionic

form

liquid

mol wt

538 g/mol

impurities

~10% water

CMC

800 ppm

HLB value

~11.7

transition temp

cloud point 36 °C

density

1.01 g/mL at 20 °C

HLB

13.1

SMILES string

O(C(CC(CC(C)C)C)CC(C)C)CCO

InChI

1S/C14H30O2/c1-11(2)8-13(5)10-14(9-12(3)4)16-7-6-15/h11-15H,6-10H2,1-5H3

InChI key

VKKFWRYCMZBXIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TERGITOL TMN 6 is a low-cost and easily available biodegradable nonionic surfactant. The hydrophobic part of this low-toxic surfactant is made up of alkyl groups and its hydrophilic part is composed of chains of oxyethylene molecules. This amphiphilic surfactant is soluble in water and polar organic solvents.

Application

TERGITOL TMN 6 is suitable for use:
  • in the slurry after ball milling to improve its wetting behavior to prepare alumina sheets
  • in the cloud point extraction process to extract organic compounds like natural estrogens E1 and E2 from urine samples
  • as an emulsifier in the optimization of the ultrasound-assisted surfactant-enhanced emulsification microextraction (UASEME) procedure to promote the extraction of benzimidazole anthelmintics
  • in the preparation of blood mimicking fluid (BMF) mixture

Biochem/physiol Actions

TERGITOL TMN 6 can be used as a synergistic cloud point extraction (SCPE) extractant. It is suitable for use in foam control, chemical mechanical planarization slurry, waterborne emulsion synthesis, and leveling.

Features and Benefits

  • Superior wetting agent
  • Serve as a penetrant & dispersant
  • Narrow gel range
  • Acts as silicone emulsifier

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alternative green preconcentration approach based on ultrasound-assisted surfactant-enhanced emulsification microextraction and HPLC for determination of benzimidazole anthelmintics in milk formulae
Boontongto T, et al.
Chromatographia, 77, 1557-1562 (2014)
Gel-tape casting as a novel method for the production of flexible fine-grained alumina sheets
Stastny P, et al.
Journal of Separation Science, 40(7), 2542-2547 (2020)
Jun Zhou et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1072, 136-141 (2017-11-21)
A novel improved preconcentration method known as synergistic cloud point extraction was established for isoquercitrin preconcentration and determination in rat plasma prior to its determination by high performance liquid chromatography. Synergistic cloud point extraction greatly simplified isoquercitrin extraction and detection.
Wall-less flow phantoms with tortuous vascular geometries: Design principles and a patient-specific model fabrication example
Ho CK, et al.
IEEE Transactions on Ultrasonics, Ferroelectrics, and Frequency Control, 64(1), 25-38 (2017)
Development and application of a cloud point method for the extraction of natural estrogens E1 and E2 from urine samples and their simultaneous determination by HPLC-EC using a BDD electrode
De Amorim KP and Andrade LS
Analytical Methods : Advancing Methods and Applications, 9(10), 1627-1633 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service