Skip to Content
Merck
All Photos(1)

Key Documents

83273

Sigma-Aldrich

Citrate Concentrated Solution

BioUltra, for molecular biology, 1 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Na3O7
CAS Number:
Molecular Weight:
258.07
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

concentration

1 M in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

7.0-8.0

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.1

SMILES string

[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;/q;3*+1/p-3

InChI key

HRXKRNGNAMMEHJ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Atanassova et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(8), 1703-1711 (2015-03-20)
Aqueous-ionic liquid (A-IL) biphasic systems have been examined in terms of deuterated water, acid, and IL cation and anion mutual solubilities in the upper (water-rich, in mole fraction) and lower phase of aqueous/IL biphasic systems at ambient temperature. The biphasic
Safiah H C Mai et al.
Shock (Augusta, Ga.), 44(2), 166-172 (2015-05-27)
Sepsis is characterized by systemic activation of coagulation and inflammation in response to microbial infection. Although cell-free DNA (cfDNA) released from activated neutrophils has antimicrobial properties, it may also exert harmful effects by activating coagulation and inflammation. The authors aimed
Kirsten Balvers et al.
Shock (Augusta, Ga.), 43(4), 317-321 (2015-01-08)
Severe trauma affects the immune system, which in its turn is associated with poor outcome. The mediators driving the immune responses in trauma are largely unknown. The aim of this study was to investigate the role of endogenous microparticles (MPs)
Katherine G MacDonald et al.
The Journal of allergy and clinical immunology, 135(4), 946--946- (2015-02-14)
Systemic sclerosis (SSc) is an autoimmune disorder characterized by fibrosis of the skin and internal organs. Pathologic conversion of regulatory T (Treg) cells into inflammatory cytokine-producing cells is thought to be an important step in the progression of autoimmunity, but
Maria T Bahia et al.
Antimicrobial agents and chemotherapy, 58(8), 4362-4370 (2014-05-21)
This study was designed to verify the in vivo efficacy of sulfoxide and sulfone fexinidazole metabolites following oral administration in a murine model of Chagas disease. Female Swiss mice infected with the Y strain of Trypanosoma cruzi were treated orally

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service