Skip to Content
Merck
All Photos(1)

Documents

81255

Sigma-Aldrich

Poly(ethylene glycol)

BioUltra, 6,000

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

form

solid

mol wt

Mr 5000-7000

impurities

insoluble matter, passes filter test
≤20 mg/kg peroxides (as H2O2)

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

solubility

H2O: 50 mg/mL at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: 0.15
λ: 280 nm Amax: 0.10

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, which is required for the purification of biological materials. PEG also interacts with cell membrane, thereby allowing cell fusion.

Application

Poly(ethylene glycol) has been used for stress (dehydration) treatment in seedlings of Galegae orientalis. It has been used for the preparation of aqueous two-phase system to study protein diffusion across the interface.

Other Notes

Fractional precipitation of proteins; Protoplast fusion; In virus purification; Electroblotting of proteins on agarose gel containing PEG and antibodies

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid bacteriophage sedimentation in the presence of polyethylene glycol and its application to large-scale virus purification.
K R Yamamoto et al.
Virology, 40(3), 734-744 (1970-03-01)
Protein diffusion across the interface in aqueous two-phase systems.
Munchow G, et al.
Langmuir, 24, 8547-8553 (2008)
K C Ingham
Methods in enzymology, 182, 301-306 (1990-01-01)
Polyethylene glycol is a nondenaturing water-soluble polymer whose ability to precipitate protein from aqueous solution can be qualitatively understood in terms of an excluded volume mechanism. The increment in PEG concentration required to effect a given reduction in solubility is
J.B. Power et al.
Methods in Enzymology, 118, 578-578 (1986)
José A Manso et al.
Structure (London, England : 1993), 27(6), 952-964 (2019-04-23)
Mechanical stability of epithelia requires firm attachment to the basement membrane via hemidesmosomes. Dysfunction of hemidesmosomal proteins causes severe skin-blistering diseases. Two plakins, plectin and BP230 (BPAG1e), link the integrin α6β4 to intermediate filaments in epidermal hemidesmosomes. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service