811D-250
Human Adipocyte Differentiation Medium (250 ml)
Synonym(s):
Primary Cell Culture Media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
Quality Level
shelf life
6 mo.
manufacturer/tradename
Cell Applications, Inc
shipped in
wet ice
storage temp.
2-8°C
General description
Human Adipocyte Differentiation Medium
Application
Human Adipocyte Differentiation Medium (250 ml) has been used to induce adipogenic differentiation in human pre-adipocytes.(14}
Quality
Each lot was tested for the ability to support plating, spreading and proliferation
Preparation Note
contained in protocol
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Oral surgery, oral medicine, oral pathology, oral radiology, and endodontics, 111(6), 700-708 (2010-12-15)
Stem cells isolated from human dental follicles as a potential cell source for bone-tissue engineering were examined for correcting a critical bone defect. Impacted third molars were collected and single cell-derived cell populations were cultivated in growth medium. Single cell-derived
Lab on a chip, 19(2), 241-253 (2018-12-20)
Infiltration of immune cells into adipose tissue is associated with chronic low-grade inflammation in obese individuals. To better understand the crosstalk between immune cells and adipocytes, in vivo-like in vitro models are required. Conventionally transwell culture plates are used for
Molecular and cellular endocrinology, 506, 110744-110744 (2020-02-07)
Triiodothyronine (T3) and irisin (I) can modulate metabolic status, increase heat production, and promote differentiation of white adipose tissue (WAT) into brown adipose tissue (BAT). Herein, human subcutaneous white adipocytes were treated with 10 nM T3 or 20 nM I for 24 h
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service