Skip to Content
Merck
All Photos(1)

Key Documents

76440

Sigma-Aldrich

Pentacene

for fluorescence, >95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912418
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

grade

for fluorescence

Quality Level

Assay

>95.0%

form

solid

mp

372-374 °C (subl.)

SMILES string

c1ccc2cc3cc4cc5ccccc5cc4cc3cc2c1

InChI

1S/C22H14/c1-2-6-16-10-20-14-22-12-18-8-4-3-7-17(18)11-21(22)13-19(20)9-15(16)5-1/h1-14H

InChI key

SLIUAWYAILUBJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maor Asher et al.
Advanced materials (Deerfield Beach, Fla.), 32(10), e1908028-e1908028 (2020-02-01)
The intermolecular lattice vibrations in small-molecule organic semiconductors have a strong impact on their functional properties. Existing models treat the lattice vibrations within the harmonic approximation. In this work, polarization-orientation (PO) Raman measurements are used to monitor the temperature-evolution of
Beomjin Park et al.
ACS nano, 9(8), 8352-8360 (2015-07-07)
Precise graphene patterning is of critical importance for tailor-made and sophisticated two-dimensional nanoelectronic and optical devices. However, graphene-based heterostructures have been grown by delicate multistep chemical vapor deposition methods, limiting preparation of versatile heterostructures. Here, we report one-pot synthesis of
Yuseong Gim et al.
Nanoscale, 7(33), 14100-14108 (2015-08-06)
Atomically-thin molecular layers of aryl-functionalized graphene oxides (GOs) were used to modify the surface characteristics of source-drain electrodes to improve the performances of organic field-effect transistor (OFET) devices. The GOs were functionalized with various aryl diazonium salts, including 4-nitroaniline, 4-fluoroaniline
Tatjana Djuric et al.
Crystal growth & design, 11(4), 1015-1020 (2011-04-12)
This study investigates epitaxially oriented pentacene films grown on Cu(110) surfaces crystallizing either in the "thin film" phase with standing molecules or in the "single crystal" structure with molecules lying with their long axes parallel to the substrate.
Jonathan Breeze et al.
Nature communications, 6, 6215-6215 (2015-02-24)
Recently, the world's first room-temperature maser was demonstrated. The maser consisted of a sapphire ring housing a crystal of pentacene-doped p-terphenyl, pumped by a pulsed rhodamine-dye laser. Stimulated emission of microwaves was aided by the high quality factor and small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service