Skip to Content
Merck
All Photos(1)

Key Documents

74283

Sigma-Aldrich

(±)-α-Carotene

≥95.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95.0% (HPLC)

form

powder

λ

2 % in methylene chloride (in hexane)

UV absorption

λ: 445-449 nm Amax

storage temp.

−20°C

SMILES string

CC(/C=C/C1C(C)=CCCC1(C)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(\C=C\C2=C(C)CCCC2(C)C)C

InChI

1S/C40H56/c1-31(19-13-21-33(3)25-27-37-35(5)23-15-29-39(37,7)8)17-11-12-18-32(2)20-14-22-34(4)26-28-38-36(6)24-16-30-40(38,9)10/h11-14,17-23,25-28,37H,15-16,24,29-30H2,1-10H3/b12-11+,19-13+,20-14+,27-25+,28-26+,31-17+,32-18+,33-21+,34-22+

InChI key

ANVAOWXLWRTKGA-JLTXGRSLSA-N

General description

α-Carotene is usually associated with β-carotene in many fruits and vegetables and belongs to the carotenoid family. It is found in higher concentrations of orange carrots, pumpkins, sweet potatoes, and apricots.

Biochem/physiol Actions

α-Carotene exhibits provitamin A (retinal/retinol/retinoic acid) activity that is necessary for enhancing ocular health. It also shows anti-oxidant, and anti-cancer activities and promotes the function of the immune system. α-Carotene has been studied as an important factor in reducing the risk of cardiovascular diseases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Carlos E Harjes et al.
Science (New York, N.Y.), 319(5861), 330-333 (2008-01-19)
Dietary vitamin A deficiency causes eye disease in 40 million children each year and places 140 to 250 million at risk for health disorders. Many children in sub-Saharan Africa subsist on maize-based diets. Maize displays considerable natural variation for carotenoid
P Rinaldi et al.
Neurobiology of aging, 24(7), 915-919 (2003-08-21)
In order to assess peripheral levels and activities of a broad spectrum of non-enzymatic and enzymatic antioxidants in elderly subjects with mild cognitive impairment (MCI) and Alzheimer's disease (AD), plasma levels of water-soluble (Vitamin C and uric acid) and of
Michael Englert et al.
Journal of chromatography. A, 1388, 119-125 (2015-03-03)
A carotenoid purification method with dual-mode countercurrent chromatography (CCC) for β-carotene, α-carotene and lutein from a fresh carrot extract was developed. The fluorinated liquid benzotrifluoride (IUPAC name: (trifluoromethyl)benzene) was used as a novel modifier in the non-aqueous ternary solvent system
N J Miller et al.
FEBS letters, 384(3), 240-242 (1996-04-22)
The purpose of this study was to assess the relative antioxidant activities of a range of carotenes and xanthophylls through the extent of their abilities to scavenge the ABTS(.+) radical cation. The results show that the relative abilities of the
Metabolism of alpha-carotene.
J S McAnally et al.
Nature, 210(5043), 1366-1366 (1966-06-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service