Skip to Content
Merck
All Photos(1)

Key Documents

72715

Sigma-Aldrich

Glutaryl-L-carnitine lithium salt

≥98.0% (TLC)

Synonym(s):

L-Carnitine lithium glutarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO6 · xLi+
CAS Number:
Molecular Weight:
275.30 (free acid basis)
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Product Name

Glutaryl-L-carnitine lithium salt, ≥98.0% (TLC)

Quality Level

Assay

≥98.0% (TLC)

form

solid

optical activity

[α]/D -20±2°, c = 1 in H2O

impurities

≤10% water

color

white to off-white

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCC(O)=O)=O)CC([O-])=O.C

InChI

1S/C12H21NO6/c1-13(2,3)8-9(7-11(16)17)19-12(18)6-4-5-10(14)15/h9H,4-8H2,1-3H3,(H-,14,15,16,17)/t9-/m1/s1

InChI key

NXJAXUYOQLTISD-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Increased formation and excretion of glutarylcarnitin results from a glutaryl-CoA dehydrogenase deficiency, an inborn error of lysine and tryptophan metabolism. Secondary carnitine depletion due to increased formation and urinary excretion of glutarylcarnitine is suggested to play an important role in the neuropathogenesis of glutaryl-CoA dehydrogenase deficiency, inducing excitotoxic neuronal damage and mitochondrial dysfunction.

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee Su Kim et al.
Annals of clinical and laboratory science, 44(2), 213-216 (2014-05-06)
Mutations in the glutaryl-CoA dehydrogenase gene can result in Glutaric aciduria type 1(GA 1) by accumulation of glutaric acid, 3-hydroxyglutaric acid (3-OH-GA), and glutarylcarnitine (C5DC). GA 1 is characterized by macrocephaly, subdural hemorrhage (SDH), and dystonic movement disorder after acute
Glutaric aciduria; a "new" disorder of amino acid metabolism.
S I Goodman et al.
Biochemical medicine, 12(1), 12-21 (1975-01-01)
Chee-Seng Lee et al.
Metabolic brain disease, 28(1), 61-67 (2012-10-30)
Glutaric aciduria type I (GA-I) is an inborn error of lysine and tryptophan metabolism. Clinical manifestations of GA-I include dystonic or dyskinetic cerebral palsy, but when the symptoms occur, treatment is not effective. In Taiwan, newborn screening for GA-I started
Kim M Huffman et al.
Diabetes care, 32(9), 1678-1683 (2009-06-09)
To determine whether circulating metabolic intermediates are related to insulin resistance and beta-cell dysfunction in individuals at risk for type 2 diabetes. In 73 sedentary, overweight to obese, dyslipidemic individuals, insulin action was derived from a frequently sampled intravenous glucose
K Kidouchi et al.
Clinica chimica acta; international journal of clinical chemistry, 164(3), 261-266 (1987-05-15)
A technique for the identification of glutarylcarnitine in urine from a patient with glutaric aciduria type 1 is described. The patient's urine sample was partially purified using an anion exchange column and analyzed by a carboxylic acid analyzer fitted with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service