Skip to Content
Merck
All Photos(1)

Key Documents

64319

Sigma-Aldrich

L-Methionine

≥99.5% (NT), BioUltra

Synonym(s):

(S)-2-Amino-4-(methylmercapto)butyric acid, L-2-Amino-4-(methylthio)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
Beilstein:
1722294
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Methionine, BioUltra, ≥99.5% (NT)

biological source

synthetic

Quality Level

product line

BioUltra

Assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D +23.7±0.5°, c = 5% in 5 M HCl

impurities

insoluble matter, passes filter test
≤0.5% foreign amino acids

ign. residue

≤0.1%

loss

≤0.1% loss on drying, 20 °C (HV)

color

white

mp

284 °C (dec.) (lit.)

solubility

1 M HCl: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤100 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 0.40
λ: 280 nm Amax: 0.05

SMILES string

CSCC[C@H](N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1

InChI key

FFEARJCKVFRZRR-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>ROS-Responsive and Self-Tumor Curing Methionine Polymer Library Based Nanoparticles with Self-Accelerated Drug Release and Hydrophobicity/Hydrophilicity Switching Capability for Enhanced Cancer Therapy</strong>: This publication details the development of L-Methionine based nanoparticles, showcasing its pivotal role in advancing drug delivery systems for cancer treatment. The study highlights the potential of these systems in enhancing therapeutic efficacy through controlled drug release (Liu et al., 2024).</li>
<li><strong>N-acetyl-l-methionine dietary supplementation improves meat quality by oxidative stability of finishing Angus heifers</strong>: This study explores the benefits of L-Methionine supplementation in animal diets, emphasizing its impact on improving meat quality and oxidative stability, which is crucial for the meat production industry and nutritional science (Liu et al., 2024).</li>
</ul>

Other Notes

Synthesis of labels for protonation studies by Fourier Transform Infrared Methods for L-glutaric-513C acid; Spacer in isotachophoresis of serum proteins; Essential in culture media for some malignant human cell lines; Growth requirement of various microorganisms

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Handbook of Microbiology, 4, 1-1 (1974)
W Kreis et al.
Cancer research, 40(3), 634-641 (1980-03-01)
Human embryonic lung fibroblasts (F-136-35-56) capable of growing in medium containing DL-homocysteine instead of L-methionine and human acute lymphoblastic leukemia cells (CCRF-HSB-2) with absolute methionine requirement exhibited dose-dependent growth inhibition when semipurified L-methionine-degrading enzyme (L-methioninase, EC 4.4.1.11) was added to
W Kreis
Cancer treatment reports, 63(6), 1069-1072 (1979-06-01)
Published reports indicate that normal rodent cells can grow in medium containing either L-methionine or L-homocysteine, whereas malignant rodent cells have an absolute requirement for L-methionine. Our studies with two normal human cell lines (fetal lung fibroblasts and bladder epithelial
T. Hine et al.
Electrophoresis ?83, 541-541 (1984)
Bacteriorhodopsin: fourier transform infrared methods for studies of protonation of carboxyl groups.
G Dollinger et al.
Methods in enzymology, 127, 649-662 (1986-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service