Skip to Content
Merck
All Photos(1)

Key Documents

63590

Sigma-Aldrich

May-Grünwald solution

Synonym(s):

Eosin – methylene blue solution, Eosin methylene blue according to May-Grünwald

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.25

technique(s)

microbe id | staining: suitable

Quality Level

impurities

methanol

density

0.80 g/mL at 20 °C

suitability

Chlamydia spp.
Histoplasma spp.
Plasmodium spp.
Trichomonas spp.
bacteria
protozoa
spirochetes

SMILES string

[K+].COC(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C21H10Br4O5.K/c1-29-21(28)9-5-3-2-4-8(9)14-10-6-12(22)17(26)15(24)19(10)30-20-11(14)7-13(23)18(27)16(20)25;/h2-7,26H,1H3;/q;+1/p-1

InChI key

NXBZXYJXNIFCRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May-Grünwald solution (Eosin – methylene blue solution) is used with Giemsa stain for histological staining of blood cells.

Other Notes

Stain for the panoptic staining according to Pappenheim

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farshad Naghshvar et al.
Cell biochemistry and function, 30(7), 569-573 (2012-04-27)
The protective effects of carnosine as a natural dipeptide were investigated in mouse bone marrow cells against genotoxicity induced by cyclophosphamide. Mice were injected with solutions of carnosine at three different doses (10, 50 and 100 mg kg(-1) bw) for
V Hofman et al.
Cytopathology : official journal of the British Society for Clinical Cytology, 23(1), 30-38 (2011-01-08)
Recurrence rates after surgery for non-small cell lung cancer (NSCLC) range from 25 to 50% and 5-year survival is only 60-70%. Because no biomarkers are predictive of recurrence or the onset of metastasis, pathological TNM (pTNM) staging is currently the
P. Bock, ed.
Romeis Mikroskopische Technik null
Tobias Koeniger et al.
Stem cells (Dayton, Ohio), 39(2), 227-239 (2020-12-04)
Although the bone marrow contains most hematopoietic activity during adulthood, hematopoietic stem and progenitor cells can be recovered from various extramedullary sites. Cells with hematopoietic progenitor properties have even been reported in the adult brain under steady-state conditions, but their
Elsa Beraki et al.
CytoJournal, 9, 8-8 (2012-05-11)
Protocols for immunocytochemical staining (ICC) and in situ hybridization (ISH) of air-dried Diff-Quick or May-Grünwald Giemsa (MGG)-stained smears have been difficult to establish. An increasing need to be able to use prestained slides for ICC and ISH in specific cases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service