Skip to Content
Merck
All Photos(1)

Key Documents

62574

Sigma-Aldrich

Lithium nitrate

BioUltra, ≥99.0% (calc. on dried substances, T)

Synonym(s):

LiNO3, Lithium salt of nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNO3
CAS Number:
Molecular Weight:
68.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent grade
form:
solid
solubility:
H2O: 1 M at 20 °C, clear, colorless

grade

reagent grade

product line

BioUltra

Assay

≥99.0% (calc. on dried substances, T)

form

solid

impurities

insoluble matter, passes filter test

loss

≤2% loss on drying, 110 °C

pH

6-9 (25 °C, 1 M in H2O)

mp

264 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤10 mg/kg
As: ≤0.1 mg/kg
Ba: ≤10 mg/kg
Bi: ≤5 mg/kg
Ca: ≤100 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 329 nm in H2O at 1 M

SMILES string

[Li+].[O-][N+]([O-])=O

InChI

1S/Li.NO3/c;2-1(3)4/q+1;-1

InChI key

IIPYXGDZVMZOAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Revealing Lithium Nitrate-Mediated Solid-Electrolyte Interphase of Lithium Metal Anode via Cryogenic Transmission Electron Microscopy.: This study explores the role of lithium nitrate in forming the solid-electrolyte interphase (SEI) on lithium metal anodes. Using cryogenic transmission electron microscopy, the researchers revealed detailed structural and compositional insights, enhancing the understanding of lithium metal batteries (Zhen et al., 2024).

  • Three-dimensional cross-linked network deep eutectic gel polymer electrolyte with the self-healing ability enabled by hydrogen bonds and dynamic disulfide bonds.: This research presents a novel gel polymer electrolyte incorporating lithium nitrate, which facilitates a self-healing mechanism through hydrogen bonds and dynamic disulfide bonds, improving battery performance and longevity (Chen et al., 2024).

  • Additive-Driven Nanoscale Architecture of Solid Electrolyte Interphase Revealed by Cryogenic Transmission Electron Microscopy.: The study highlights how lithium nitrate, as an additive, influences the nanoscale architecture of the SEI, providing crucial insights for the design of better lithium-ion batteries (Park et al., 2024).

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syed Nasir Abbas Bukhari et al.
Mini reviews in medicinal chemistry, 12(13), 1394-1403 (2012-08-11)
Chalcones are the principal precursors for the biosynthesis of flavonoids and isoflavonoids. A three carbon α, β-unsaturated carbonyl system constitutes chalcones. Chalcones are the condensation products of aromatic aldehyde with acetophenones in attendance of catalyst. They go through an assortment
M Esthaku Peter et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(5), 1417-1421 (2010-03-20)
Single crystal of gamma-glycine, an organic nonlinear optical material, has been grown by solvent evaporation technique from a mixture of aqueous solutions of glycine and potassium nitrate, lithium nitrate at room temperature. Gamma glycine crystals have been grown up to
Anastassios C Papageorgiou et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 62(Pt 3), 242-244 (2006-03-03)
Superantigens are bacterial or viral toxins with potent immunostimulatory properties. Streptococcus dysgalactiae-derived mitogen, a 25 kDa protein, is a recently discovered superantigen isolated from S. dysgalactiae culture supernatant. Sequence considerations suggest that it belongs to a new superantigen family distinct
Som Datta Sharma et al.
Biological & pharmaceutical bulletin, 28(5), 834-837 (2005-05-03)
Lithium (Li(+)) salts are commonly used in treating bipolar diseases. As physicians frequently keep the patients on long-term lithium therapy, awareness of the numerous side effects and pathogenesis of this lightest alkali metal is needed for such treatments. The current
Xingwen Yu et al.
ACS applied materials & interfaces, 7(30), 16625-16631 (2015-07-15)
Lithium-sulfur (Li-S) batteries are receiving great attention as the most promising next-generation power source with significantly high charge-storage capacity. However, the implementation of Li-S batteries is hampered by a critical challenge because of the soluble nature of the intermediate polysulfide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service