Skip to Content
Merck
All Photos(1)

Key Documents

62335

Sigma-Aldrich

Lipoprotein Lipase from Pseudomonas sp.

lyophilized, powder, ≥1200 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Pseudomonas spp.)

Quality Level

form

powder

quality

lyophilized

specific activity

≥1200 U/mg

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Lipoprotein Lipase (LPL) is majorly secreted by myocytes and adipocytes in humans and is crucial for triglyceride homeostatis. Mutations in the catalytic domain of LPL impairs its interaction with glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1 (GPIHBP1). The N-terminal catalytic domain is essential for lipolysis. The C-terminal is crucial for binding lipoproteins. Altered LPL levels may play role in the pathogenesis of atherosclerosis, coronary heart disease and chronic lymphocytic leukemia.

Application

Lipoprotein Lipase from Pseudomonas sp. has been used in the enzymatic erosion studies in gamma irradiated poly(trimethylene carbonate) (PTMC) films.

Biochem/physiol Actions

Lipoprotein lipase belongs to the family of triglyceride lipases. It hydrolyses triglycerides in triglyceride-rich ApoB-containing lipoproteins.
Malic dehydrogenase catalyzes the dehydrogenation of L-malate by NAD+.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40°C (triolein, Cat. No. 62314 as substrate)

Other Notes

Preparation of aldol acceptors (R)- and (S)-3-azido-2-hydroxypropanal via lipase-catalyzed resolution of the racemic acetal precursorcitation; Effect of enzyme form on its properties in toluenecitation
Preparation of aldol acceptors (R)- and (S)-3-azido-2-hydroxypropanal via lipase-catalyzed resolution of the racemic acetal precursor; Effect of enzyme form on its properties in toluene

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.H. von der Osten et al.
Journal of the American Chemical Society, 111, 3924-3924 (1989)
Mutations in lipoprotein lipase that block binding to the endothelial cell transporter GPIHBP1
Voss CV, et al.
Proceedings of the National Academy of Sciences of the USA, 108(19), 7980-7984 (2011)
A lipoprotein lipase (LPL)-specific monoclonal antibody, 88B8, that abolishes the binding of LPL to GPIHBP1
Allan CM, et al.
Journal of Lipid Research, jlr-M070813 (2016)
G. Ottolina et al.
Biotechnology Letters, 14, 947-947 (1992)
Macrophage-mediated erosion of gamma irradiated poly (trimethylene carbonate) films
Bat E, et al.
Biomaterials, 30(22), 3652-3661 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service