Skip to Content
Merck
All Photos(1)

Documents

60359

Sigma-Aldrich

Potassium phthalate monobasic

BioUltra, ≥99.5% (T)

Synonym(s):

KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.21

product line

BioUltra

Assay

≥99.5% (T)

form

powder or crystals

impurities

insoluble matter, passes filter test

loss

≤0.05% loss on drying, 110 °C

pH

3.5-5.0 (25 °C, 0.1 M in H2O)

mp

295-300 °C (dec.) (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 309 nm in H2O at 0.1 M

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium hydrogen phthalate may be used as a buffering agent or as a standard for total organic carbon (TOC) calibration and may be used to asses the oxidating activity of CNTs/TiO(2) photoanodes. Potassium hydrogen phthalate solution may be used as a liquid filter to reduce the effects of Rayleigh scattering on collected Raman spectra.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E A Saunders et al.
The Review of scientific instruments, 81(1), 013108-013108 (2010-02-02)
Broadband XeCl excimer lasers operating at 308 nm are not currently used in the field of gas phase vibrational Raman spectroscopy (VRS). An explanation as to why alternative wavelengths, and in particular tuneable, narrowband lasers are currently preferred for gas
Maria Cristina Fossi et al.
Marine pollution bulletin, 64(11), 2374-2379 (2012-09-12)
Baleen whales are potentially exposed to micro-litter ingestion as a result of their filter-feeding activity. However, the impacts of microplastics on baleen whales are largely unknown. In this case study of the Mediterranean fin whale (Balaenoptera physalus), we explore the
Juanjuan Feng et al.
Journal of chromatography. A, 1268, 16-21 (2012-11-07)
Because of the occurrence of ion exchange between high-ionic-strength solution and anions of polymeric ionic liquids (PILs), PILs based solid-phase microextraction (SPME) fibers were rarely used in direct immersion mode to high-salt-added samples. In this work, a novel double-confined PIL
Hanne Frederiksen et al.
Environmental science & technology, 47(2), 958-967 (2012-12-14)
Urinary phthalate excretion is used as marker of phthalate exposure in epidemiological studies. Here we examine the reliability of urinary phthalate levels in exposure classification by comparing the inter- and intrasubject variation of urinary phthalate metabolite levels. Thirty-three young healthy
Randi J Bertelsen et al.
Environmental health perspectives, 121(2), 251-256 (2012-11-21)
High-molecular-weight phthalates in indoor dust have been associated with asthma in children, but few studies have evaluated phthalate biomarkers in association with respiratory outcomes. We explored the association between urinary concentrations of phthalate metabolites and current asthma. In a cross-sectional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service