Skip to Content
Merck
All Photos(1)

Key Documents

53489

Sigma-Aldrich

Callistephin chloride

≥95.0% (HPLC)

Synonym(s):

3-(Glucosyloxy)-4′,5,7-trihydroxyflavylium chloride, Pelargonidin 3-O-glucoside chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClO10
CAS Number:
Molecular Weight:
468.84
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥95.0% (HPLC)

form

powder

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O)cc(O)cc3[o+]c2-c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O10.ClH/c22-8-16-17(26)18(27)19(28)21(31-16)30-15-7-12-13(25)5-11(24)6-14(12)29-20(15)9-1-3-10(23)4-2-9;/h1-7,16-19,21-22,26-28H,8H2,(H2-,23,24,25);1H/t16-,17-,18+,19-,21-;/m1./s1

InChI key

CAHGSEFWVUVGGL-UBNZBFALSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Callistephin chloride is an anthocyanin which can be found in berries, for example in strawberries and chokeberries. Is has antioxidant activity[1] and is examined for its neuroprotective properties[2].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Natalie Kelsey et al.
Nutritional neuroscience, 14(6), 249-259 (2011-11-08)
Mitochondrial oxidative stress (MOS) is a major factor in the underlying pathology of many neurodegenerative diseases. Here, we investigated the neuroprotective effects of a unique class of nutraceutical antioxidants, anthocyanins, against MOS-induced death of cultured cerebellar granule neurons (CGNs). Callistephin
W Nerdal et al.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989), 46(9), 872-876 (1992-09-01)
Pelargonidin-3-glucoside has been isolated from the acidified methanolic extract of strawberries (Fragaria anannassa variety Corona) by successive application of an ion-exchange resin, droplet-counter chromatography and gel filtration. The pigment in acidified methanolic solution was studied by means of the two-dimensional
I Zabetakis et al.
Journal of agricultural and food chemistry, 48(7), 2749-2754 (2000-07-18)
Color stability of fruit juice made from strawberries (Fragaria x ananassa, cv. Elsanta) that were subjected to high hydrostatic pressure was studied by measuring the anthocyanin content. High hysrostatic pressure is a method of preservation of food alternative to heat
Colleen Carkeet et al.
The Journal of nutrition, 138(5), 897-902 (2008-04-22)
A clinical study was conducted to investigate the dose response and metabolism of strawberry anthocyanins. In a crossover study design, 12 healthy adults consumed each of 3 strawberry treatments. The treatments were 100 g, 200 g, and 400 g of
Maarit J Eiro et al.
Journal of agricultural and food chemistry, 50(25), 7461-7466 (2002-11-28)
Intermolecular copigmentation reactions are significantly responsible for the manifold color expression of fruits, berries, and their products. These reactions were investigated with five anthocyanins and five phenolic acids acting as copigments. The stability of the pigment-copigment complexes formed was studied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service