Skip to Content
Merck
All Photos(1)

Key Documents

51085

Sigma-Aldrich

Isobutyryl-L-carnitine

≥97.0% (TLC)

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-(2-methyl-1-oxopropoxy)-1-propanaminium inner salt, L-Carnitine isobutyryl ester, iC4-Carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO4
CAS Number:
Molecular Weight:
231.29
Beilstein:
4138517
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.26

product name

Isobutyryl-L-carnitine, ≥97.0% (HPLC)

Quality Level

Assay

≥97.0% (HPLC)

form

powder

optical activity

[α]/D -22±2°, c = 1 in H2O

impurities

≤10% water

color

white

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(C(C)C)=O)CC([O-])=O

InChI

1S/C11H21NO4/c1-8(2)11(15)16-9(6-10(13)14)7-12(3,4)5/h8-9H,6-7H2,1-5H3/t9-/m1/s1

InChI key

LRCNOZRCYBNMEP-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Isobutyryl-L-carnitine is a product of the acyl-CoA dehydrogenases (ACADs), which are a group of mitochondrial enzymes involved in the metabolism of fatty acids or branched-chain amino acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L Vallée et al.
European journal of pediatrics, 153(8), 598-603 (1994-08-01)
We describe on a 3-year-old child referred for evaluation and therapy of a cerebral vascular accident with residual hemiplegia and partial epilepsy. Metabolic investigations initially showed normal urinary organic acids as well as normal blood and urinary amino acids. Blood
N Sugiyama et al.
Acta paediatrica Japonica : Overseas edition, 32(4), 410-416 (1990-08-01)
A large quantity of propionylcarnitine in the urine of patients with propionic acidemia and methylmalonic aciduria was demonstrated. The amount excreted depended on the administered L-carnitine dose from 25 to 75 mg/kg/day. A high level of propionylcarnitine was also detected
Dwight D Koeberl et al.
Pediatric research, 54(2), 219-223 (2003-05-09)
Tandem mass spectrometry was adopted for newborn screening by North Carolina in April 1999. Since then, three infants with short-chain acyl-CoA dehydrogenase (SCAD) and one with isobutyryl-CoA dehydrogenase deficiency were detected on the basis of elevated butyrylcarnitine/isobutyrylcarnitine (C4-carnitine) concentrations in
M Fontaine et al.
Clinica chimica acta; international journal of clinical chemistry, 273(2), 161-170 (1998-07-10)
Here we report on a patient with severe ("non-classic") carnitine palmitoyltransferase type II (CPT II) deficiency. Hypoglycemia prompted by an infectious episode and associated with non-ketotic dicarboxylic aciduria orientated diagnosis towards beta-oxidation deficiency disorders. Blood carnitine levels revealed a secondary
K Matsumoto et al.
Clinica chimica acta; international journal of clinical chemistry, 216(1-2), 135-143 (1993-07-16)
An immobilized enzyme reactor, made up acylcarnitine hydrolase, carnitine dehydrogenase and diaphorase in sequence, was developed for the sensitive and selective determination of urinary free and individual acylcarnitines by a reversed-phase high-performance liquid chromatography. A 100-microliter urine sample was directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service