Skip to Content
Merck
All Photos(1)

Key Documents

50980

Sigma-Aldrich

Guanidine thiocyanate

BioUltra, for molecular biology, ≥99.0% (AT)

Synonym(s):

Guanidinium rhodanide, Guanidinium thiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
Beilstein:
3563461
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.0% (AT)

form

solid

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

ign. residue

≤0.5% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

pH

4.5-7.0 (25 °C, 6 M in H2O)

mp

117-120 °C
120-122 °C (lit.)

solubility

H2O: 6 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5000 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤5000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 280 nm in H2O at 6 M

SMILES string

SC#N.NC(N)=N

InChI

1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H

InChI key

ZJYYHGLJYGJLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chaotropic agent and strong denaturant; solubilizes cells.

Other Notes

Isolation of RNA; A new guanidine thiocyanate procedure for the isolation of full-length RNA templates for reverse transcription from tissues rich in RNase and proteoglycans

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Groppe et al.
Analytical biochemistry, 210(2), 337-343 (1993-05-01)
RNA isolated by conventional guanidinium isothiocyanate methods from tissues of a mollusc (red abalone: Haliotis rufescens) is largely degraded and discolored by contaminants. These contaminants are associated with inhibition of reverse transcriptase, prevent accurate spectrophotometric determination of RNA concentration, and
P Chomczynski et al.
Analytical biochemistry, 162(1), 156-159 (1987-04-01)
A new method of total RNA isolation by a single extraction with an acid guanidinium thiocyanate-phenol-chloroform mixture is described. The method provides a pure preparation of undegraded RNA in high yield and can be completed within 4 h. It is
Turpen, T.H. and Griffith, O.M.
Biotechniques, 4, 11-11 (1986)
Piotr Chomczynski et al.
Nature protocols, 1(2), 581-585 (2007-04-05)
Since its introduction, the 'single-step' method has become widely used for isolating total RNA from biological samples of different sources. The principle at the basis of the method is that RNA is separated from DNA after extraction with an acidic
Olesya V Stepanenko et al.
PloS one, 7(11), e48809-e48809 (2012-11-13)
Having a high folding efficiency and a low tendency to aggregate, the superfolder GFP (sfGFP) offers a unique opportunity to study the folding of proteins that have a β-barrel topology. Here, we studied the unfolding-refolding of sfGFP that was induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service