Skip to Content
Merck
All Photos(1)

Key Documents

48770

Sigma-Aldrich

Gentian violet

for microscopy (Bact., Hist.)

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
8464808
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bact., Hist.)

form

powder

density

1.19 g/cm3 at 20 °C

εmax

≥1300 at 580-595 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

General description

mixture of crystal violet [C.I.No. 42555] and methyl violet [C.I.No. 42535]

Other Notes

Dye employed for preparing Gram stain

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Bock, ed.
Romeis Mikroskopische Technik null
José Mauricio dos Santos Filho et al.
Bioorganic & medicinal chemistry, 17(18), 6682-6691 (2009-08-18)
Research in recent years has demonstrated that the Trypanosoma cruzi cysteine protease cruzain (TCC) is a valid chemotherapeutic target, since inhibitors of this protease affect the pathology appropriately. By exploring the N-acylhydrazones (NAH) as privileged structures usually present in antiparasitic
Eufrânio N da Silva Júnior et al.
Bioorganic & medicinal chemistry, 16(9), 5030-5038 (2008-04-02)
New naphthoquinone derivatives were synthesized and assayed against bloodstream trypomastigote forms of Trypanosoma cruzi, the etiological agent of Chagas' disease. The compounds were rationalized based on hybrid drugs and appear as important compounds against this parasite. From nor-lapachol were prepared
Mariana Boiani et al.
Journal of medicinal chemistry, 49(11), 3215-3224 (2006-05-26)
Three series of benzimidazole N-oxide derivatives were developed and were examined for their activity against trypanosomatid parasites (Trypanosoma cruzi and Leishmania spp.). 2H-benzimidazole 1,3-dioxides displayed remarkable in vitro activities against both parasites, with derivatives 28, 29, and 32 being the
Eufrânio N da Silva et al.
European journal of medicinal chemistry, 43(8), 1774-1780 (2007-11-30)
[1,2,3]-Triazole derivatives of nor-beta-lapachone were synthesized and assayed against the infective bloodstream trypomastigote form of Trypanosoma cruzi, the etiological agent of Chagas disease. All the derivatives were more active than the original quinones, with IC(50)/1 day values in the range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service