Skip to Content
Merck
All Photos(1)

Key Documents

45756

Supelco

Teflubenzuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H6Cl2F4N2O2
CAS Number:
Molecular Weight:
381.11
Beilstein:
8229925
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1cccc(F)c1C(=O)NC(=O)Nc2cc(Cl)c(F)c(Cl)c2F

InChI

1S/C14H6Cl2F4N2O2/c15-5-4-8(12(20)10(16)11(5)19)21-14(24)22-13(23)9-6(17)2-1-3-7(9)18/h1-4H,(H2,21,22,23,24)

InChI key

CJDWRQLODFKPEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Z I Finkelstein et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 36(5), 559-567 (2001-10-16)
Transformation of teflubenzuron, the active component in the insecticide commercialized as Nomolt, by soil microorganisms was studied. It was shown that microorganisms, belonging to Bacillus, Alcaligenes, Pseudomonas and Acinetobacter genera are capable to perform the hydrolytic cleavage of the phenylurea
Jean-Charles Bouvier et al.
Archives of insect biochemistry and physiology, 51(2), 55-66 (2002-09-17)
Insecticide resistance in the codling moth, Cydia pomonella, partly results from increased metabolic detoxification. The aim of this study was to follow the age variations in larval susceptibility to deltamethrin and teflubenzuron in one susceptible (S) strain, and two resistant
Pål A Olsvik et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 201, 35-43 (2017-09-25)
Concerns have been raised over the environmental impacts of antiparasitic drugs used to delouse farmed salmon. Released into the marine environment, some of these drugs can have negative impact on non-targeted crustaceans in the vicinity of farming facilities. In this
Fatma Acheuk et al.
Journal of insect physiology, 58(3), 335-341 (2011-12-31)
The effects of a methanolic extract of the plant Haplophyllum tuberculatum (ME-Ht) and of teflubenzuron (TFB) were compared on several reproductive variables and ecdysteroid titers in the females of Locusta migratoria. The test products were administered orally to newly emerged
A Ramstad et al.
Diseases of aquatic organisms, 50(1), 29-33 (2002-08-03)
Four commercial salmon farms on the West coast of Norway were recruited to a programme of field trials in which the efficacy of SLICE (0.2% emamectin benzoate; Schering-Plough Animal Health) was compared with a commercially available product, EKTOBANN (teflubenzuron 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service