Skip to Content
Merck
All Photos(1)

Key Documents

452M-9

Sigma-Aldrich

Cytokeratin (CAM 5.2) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

CAM 5.2, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (452M-94)
vial of 0.5 mL concentrate (452M-95)
bottle of 1.0 mL predilute (452M-97)
vial of 1.0 mL concentrate (452M-96)
bottle of 7.0 mL predilute (452M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG2aκ

control

appendix, hepatocellular carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-CAM 5.2 is a mouse monoclonal antibody that was generated by using the human colorectal carcinoma cell line HT24. Anti-CAM 5.2 reacts with cytokeratins 7 and 8, which are expressed in simple and glandular epithelia and transitional epithelium. Anti-CAM 5.2 reacts with most normal epithelium and with most carcinomas, including those arising in the lung, liver, pancreas, GI tract, breast, genitourinary system, female reproductive organs, and some endocrine organs. This antibody can be used as an aid to identify epithelial differentiation in normal and neoplastic tissues.

Linkage

Cytokeratin (CAM 5.2) Positive Control Slides, Product No. 452S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chin-Chen Pan et al.
Applied immunohistochemistry & molecular morphology : AIMM, 13(4), 347-352 (2005-11-11)
The differential diagnoses of hepatocellular carcinoma (HCC), renal cell carcinoma (RCC), and adrenocortical carcinoma (ACC) are sometimes difficult due to their overlapping histologic features. Immunohistochemistry is a helpful adjunct in supporting the histologic diagnosis. In this study, the authors used
Feiling Feng et al.
Oncotarget, 8(3), 5349-5360 (2016-12-29)
Gallbladder sarcomatoid carcinoma is a rare cancer with no clinical standard treatment. With the rapid development of next generation sequencing, it has been able to provide reasonable treatment options for patients based on genetic variations. However, most cancer drugs are
J H Sinard
Archives of ophthalmology (Chicago, Ill. : 1960), 117(6), 776-783 (1999-06-16)
Diagnosis of sebaceous carcinoma of the periorbital region is often delayed. Clinically, this lesion can mimic several inflammatory disorders. Histopathologically, it can mimic either squamous cell or basal cell carcinoma. To identify an immunohistochemical approach to assist in the diagnosis
M M Cosgrove et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 6(3), 342-347 (1993-05-01)
In this study, 29 formalin-fixed, paraffin-embedded astrocytic tumors were analyzed immunocytochemically with the antikeratin monoclonal antibodies Mak-6 and Cam 5.2 and a polyclonal antibody against glial fibrillary acidic protein (GFAP). Immunoreactivity for Mak-6 was present in 29 cases (100%) including
Nelson G Ordóñez
Human pathology, 44(7), 1195-1215 (2013-02-23)
A relatively large number of broad-spectrum immunohistochemical epithelial markers that can be used as part of the screening panels employed in the recognition of the main cell lineages during the initial evaluation of a poorly differentiated tumor are currently available.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service