Skip to Content
Merck
All Photos(1)

Key Documents

45253

Sigma-Aldrich

Wright Stain solution

for microscopy

Synonym(s):

Eosin Methylene blue solution according to Wright, Wright’s eosin methylene blue

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

impurities

methanol

density

0.80 g/mL at 20 °C

suitability

Chlamydia spp.
Histoplasma spp.
Plasmodium spp.
Trichomonas spp.
bacteria
protozoa
spirochetes

SMILES string

[K+].COC(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C21H10Br4O5.K/c1-29-21(28)9-5-3-2-4-8(9)14-10-6-12(22)17(26)15(24)19(10)30-20-11(14)7-13(23)18(27)16(20)25;/h2-7,26H,1H3;/q;+1/p-1

InChI key

NXBZXYJXNIFCRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Wright Stain is a modified Romanowsky stain commonly used for the cellular elements of blood. Wright′s stain is a mixture of eosin Y with polychromed (oxidized and partly demethylated) methylene blue and therefore termed polychrome stains. The hematological stain is intended for differentially staining peripheral blood and bone marrow films.

Application

Wright stain has been used to determine the number of white blood cell types (neutrophils, lymphocytes, monocytes, eosinophils, and basophils) per microliter of whole blood.

Other Notes

Blood stain

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Borrelia burgdorferi has minimal impact on the Lyme disease reservoir host Peromyscus leucopus.
Schwanz LE, et al.
Vector Borne and Zoonotic Diseases (Larchmont, N.Y.), 117-124 (2011)
Bernadette FR Jacqueline HC et al.
Clinical Hematology, 3rd Ed. Henry Kimpton (2015)
Analysis and testing of biological stains--the Biological Stain Commission Procedures.
Penney D P, et al.
Biotechnic & Histochemistry (2002)
K. Leybold et al.
Praxis-Laboratorium: Grundlagen der Laboratoriumsdiagnostik, 24-24 (1976)
Young-Joo Jeong et al.
Anatomy & cell biology, 43(4), 294-302 (2011-01-27)
Asthma is a Th2-dependent disease mediated by IgE and Th2 cytokines, and asthmatic patients suffer from oxidative stresses from abnormal airway inflammation. Vitamin C is a micro-nutrient functioning as an antioxidant. When administered at a mega-dose, vitamin C has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service