Skip to Content
Merck
All Photos(3)

Key Documents

445R-1

Sigma-Aldrich

NKX2.2 (EP336) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP336, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (445R-14)
vial of 0.1 mL concentrate Research Use Only (445R-14-RUO)
vial of 0.5 mL concentrate (445R-15)
vial of 1.0 mL concentrate (445R-16)
vial of 1.0 mL concentrate Research Use Only (445R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (445R-17-RUO)
vial of 1.0 mL pre-dilute ready-to-use (445R-17)
vial of 7.0 mL pre-dilute ready-to-use (445R-18)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (445R-18-RUO)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500 (concentrated)

isotype

IgG

control

Ewing’s sarcoma, pancreas

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

NKX2.2 is a homeodomain transcription factor that plays a role in neuroendocrine and glial differentiation. NKX2.2 is upregulated in Ewing′s sarcoma as a result of the oncogenic EWS-FLI1 fusion protein. As one of many small round blue cell tumors, Ewing′s sarcoma can be difficult to diagnose due to the characteristic undifferentiated morphology. NKX2.2 has proven its utility as a sensitive maker for distinguishing Ewing′s sarcoma from other round blue cell tumors when used in a panel.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Smith et al.
Cancer cell, 9(5), 405-416 (2006-05-16)
Our understanding of Ewing's sarcoma development mediated by the EWS/FLI fusion protein has been limited by a lack of knowledge regarding the tumor cell of origin. To circumvent this, we analyzed the function of EWS/FLI in Ewing's sarcoma itself. By
Arvind Rajwanshi et al.
Journal of cytology, 26(1), 1-10 (2009-01-01)
Malignant small round cell tumors are characterised by small, round, relatively undifferentiated cells. They generally include Ewing's sarcoma, peripheral neuroectodermal tumor, rhabdomyosarcoma, synovial sarcoma, non-Hodgkin's lymphoma, retinoblastoma, neuroblastoma, hepatoblastoma, and nephroblastoma or Wilms' tumor. Other differential diagnoses of small round
Yin P Hung et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 29(4), 370-380 (2016-02-06)
Ewing sarcoma shows considerable histologic overlap with other round cell tumors. NKX2-2, a homeodomain transcription factor involved in neuroendocrine/glial differentiation and a downstream target of EWSR1-FLI1, has been reported as an immunohistochemical marker for Ewing sarcoma. We assessed the specificity
Yoshinari Yamamoto et al.
Journal of cytology, 32(1), 30-32 (2015-05-08)
Renal Ewing sarcoma (ES) is a rare malignant tumor characterized by fusion of the EWSR1 gene with a member of the ETS family of oncogenes, arising at a specific chromosomal translocation. Diagnosis of ES can be problematic, especially from cytological
Akihiko Yoshida et al.
The American journal of surgical pathology, 36(7), 993-999 (2012-03-27)
Ewing sarcoma is a high-grade round cell sarcoma that affects bones and soft tissues in children and young adults. Its diagnosis can be challenging, and the differential diagnoses include a wide variety of small round cell tumors. CD99 and FLI-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service