Skip to Content
Merck

42002

Sulfo-N-succinimidyl 4-maleimidobutyrate sodium salt

~90%

Synonym(s):

4-Maleimidobutyric acid sulfo-N-succinimidyl ester sodium salt, N-(γ-Maleimidobutyryloxy)sulfosuccinimide sodium salt, Sulfo-GMBS

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C12H11N2NaO9S
CAS Number:
Molecular Weight:
382.28
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Sulfo-N-succinimidyl 4-maleimidobutyrate sodium salt, ~90%

InChI

1S/C12H12N2O9S.Na/c15-8-3-4-9(16)13(8)5-1-2-11(18)23-14-10(17)6-7(12(14)19)24(20,21)22;/h3-4,7H,1-2,5-6H2,(H,20,21,22);/q;+1/p-1

SMILES string

O=C(N1CCCC(ON2C(CC(S(=O)(O[Na])=O)C2=O)=O)=O)C=CC1=O

InChI key

ULARYIUTHAWJMU-UHFFFAOYSA-M

assay

~90%

reaction suitability

reagent type: linker

storage temp.

−20°C

Quality Level

Other Notes

Reagent for amine-to-thiol cross-linking

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Jin et al.
Colloids and surfaces. B, Biointerfaces, 136, 838-844 (2015-11-08)
In this study, elucidation of protein adsorption mechanism is performed using dual polarization interferometry (DPI) and quartz crystal microbalance with dissipation (QCM-D) to study adsorption behaviors of bovine serum albumin (BSA) and lysozyme (LYZ) on poly (ethylene glycol) (PEG) layers.
Christian NDong et al.
PloS one, 10(2), e0115636-e0115636 (2015-02-20)
Realizing the full potential of iron oxide nanoparticles (IONP) for cancer diagnosis and therapy requires selective tumor cell accumulation. Here, we report a systematic analysis of two key determinants for IONP homing to human breast cancers: (i) particle size and
Ilaria Rea et al.
Biochimica et biophysica acta, 1840(12), 3393-3403 (2014-09-17)
Diatomite is a natural porous biomaterial of sedimentary origin, formed by fragments of diatom siliceous skeletons, called "frustules". Due to large availability in many areas of the world, chemical stability, and non-toxicity, these fossil structures have been widespread used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service