Skip to Content
Merck
All Photos(1)

Key Documents

41784

Sigma-Aldrich

Atto 647 maleimide

BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (coupling to thiols)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 647 nm; λem 669 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

Application

Atto 647 belongs to a new generation of fluorescent labels for the red spectral region with high molecular absorption (120.000) and quantum yield (0.20) as well as sufficient stoke′s shift (excitation maximum 645 nm, emission maximum 669 nm). Atto 647 is characterized by a high photostability. The dye is designed for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Atto The dye is very stable under physiological pH-values and in buffers up to pH 8.5. However, it slowly degrades at higher pH. Atto 647 maleimide shows excellent solubility in polar solvents like DMF, DMSO or acetonitrile. After coupling to a substrate the dye moiety is electrically neutral.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Puntener et al.
Journal of virology, 85(1), 481-496 (2010-11-05)
Human adenoviruses (Ads) replicate and assemble particles in the nucleus. They organize a linear double-strand DNA genome into a condensed core with about 180 nucleosomes, by the viral proteins VII (pVII), pX, and pV attaching the DNA to the capsid.
Jean-Manuel Segura et al.
The Journal of biological chemistry, 283(35), 24254-24263 (2008-06-27)
CD8(+) cytotoxic T lymphocytes (CTL) can recognize and kill target cells expressing only a few cognate major histocompatibility complex (MHC) I-peptide complexes. This high sensitivity requires efficient scanning of a vast number of highly diverse MHC I-peptide complexes by the
Duncan Hill et al.
Biomedical microdevices, 13(4), 759-767 (2011-05-12)
This paper presents an overview of development of a novel disposable plastic biochip for multiplexed clinical diagnostic applications. The disposable biochip is manufactured using a low-cost, rapid turn- around injection moulding process and consists of nine parabolic elements on a
Thorben Cordes et al.
Physical chemistry chemical physics : PCCP, 13(14), 6699-6709 (2011-02-12)
Modern fluorescence microscopy applications go along with increasing demands for the employed fluorescent dyes. In this work, we compared antifading formulae utilizing a recently developed reducing and oxidizing system (ROXS) with commercial antifading agents. To systematically test fluorophore performance in
Laura Pastorino et al.
Journal of colloid and interface science, 357(1), 56-62 (2011-02-26)
The protein collagen is the major component of connective tissue and it is involved in many biological functions. Its degradation is at the basis of different pathological processes. The up-regulated expression of matrix metalloproteinases and the down-regulated expression of their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service