Skip to Content
Merck
All Photos(1)

Key Documents

415M-1

Sigma-Aldrich

EZH2 (11) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

11, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (415M-14)
vial of 0.5 mL concentrate (415M-15)
bottle of 1.0 mL predilute (415M-17)
vial of 1.0 mL concentrate (415M-16)
bottle of 7.0 mL predilute (415M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

breast carcinoma, prostate adenocarcinoma, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... EZH2(2146)

General description

Enhancer of zeste homolog 2 (EZH2) is the catalytic subunit of polycomb repressive complex 2 (PRC2). It generates a methylation epigenetic mark at lysine 27 residue of histone H3 (H3K27me3) in order to silence gene expression. EZH2 target genes are involved in a variety of biological processes such as stem cell pluripotency, cell proliferation, and oncogenic transformation. EZH2 is frequently overexpressed in many cancer types. Hyperactivation of EZH2, either by overexpression or muta­tions, is found in a variety of malignancies including prostate, breast, uterine, gastric, and renal cell cancers in addition to melanoma. EZH2 overexpression has been reported in non-small cell lung cancers and lymphoma.The EZH2 protein is rarely detected in normal breast duct epithelium and in normal and hyperplastic lymph node. EZH2 is usually expressed in follicular centers, but not in mantle zones, follicular and interfollicular T cells, plasma cells or NK/T cells. However, its expression can be seen in most B-cell and T-cell lymphomas, although only in 20% (1/5) of small lymphocytic lymphoma and 14% (1/7) of plasma cell myeloma. Plasmacytoma, lymphoplasmacytic lymphoma, and MALT lymphoma have not been shown to express this protein. Recent studies also have demonstrated EZH2 is aberrantly over-expressed in pediatric rhabdomyosarcoma, independent of the histological subtypes. In summary, EZH2 correlates with tumor proliferation and may be used in an antibody panel to differentiate proliferative/aggressive lymphoma variants from indolent ones and normal resting cell populations

Quality


IVD

IVD

IVD

RUO

Linkage

EZH2 Positive Control Slides, Product No. 415S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lamia Abd Al Kader et al.
Virchows Archiv : an international journal of pathology, 463(5), 697-711 (2013-08-21)
Polycomb group (PcG) proteins are important for the regulation of hematopoiesis by regulating chromatin compaction and silencing genes related to differentiation and cell cycle. Overexpression of enhancer of zeste homologue 2 (Ezh2) and Bmi-1/PCGF4 has been implicated in solid organ
Jin-zhi Tan et al.
Acta pharmacologica Sinica, 35(2), 161-174 (2013-12-24)
EZH2 is the catalytic subunit of the polycomb repressive complex 2 (PRC2), which is a highly conserved histone methyltransferase that methylates lysine 27 of histone 3. Overexpression of EZH2 has been found in a wide range of cancers, including those
Roberta Ciarapica et al.
BMC medicine, 9, 63-63 (2011-05-26)
Soft tissue sarcomas of childhood are a group of heterogeneous tumors thought to be derived from mesenchymal stem cells. Surgical resection is effective only in about 50% of cases and resistance to conventional chemotherapy is often responsible for treatment failure.
Sooryanarayana Varambally et al.
Nature, 419(6907), 624-629 (2002-10-11)
Prostate cancer is a leading cause of cancer-related death in males and is second only to lung cancer. Although effective surgical and radiation treatments exist for clinically localized prostate cancer, metastatic prostate cancer remains essentially incurable. Here we show, through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service