Skip to Content
Merck
All Photos(1)

Documents

40839

Sigma-Aldrich

Anti-Rabbit-IgG - Atto 647N antibody produced in goat

1 mg/mL IgG

Synonym(s):

Atto 647N-Anti-Rabbit-IgG antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

Atto 647N conjugate

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

50% glycerol as stabilizer

species reactivity

rabbit

concentration

1 mg/mL IgG

technique(s)

immunofluorescence: suitable (5μg/ml)

fluorescence

λex 647 nm; λem 665 nm in PBS

suitability

in accordance for fluorescence

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Goat anti-rabbit IgGs are known to associate with rabbit IgGs.

Immunogen

rabbit IgG

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation.Atto 647N goat anti-rabbit IgG was used as the secondary antibody for immunofluorescene at a concentration of 5μg/ml on cells fixed in 2% formaldehyde.

Physical form

Atto 647 goat anti-rabbit IgG (whole molecule) is provided in unit sizes of 1 ml as 1 mg/ml solution in 0.1 M sodium phosphate, 0.1 M NaCl, pH 7.5, containing 5 mM sodium azide as a preservative.

Analysis Note

unconjugated dye ≤5% of total fluorescence

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masfique Mehedi et al.
Bio-protocol, 7(17) (2017-10-24)
Human respiratory syncytial virus (RSV) infection in human lung epithelial A549 cells induces filopodia, cellular protrusions consisting of F-actin, that extend to neighboring uninfected cells (Mehedi et al., 2016). High-resolution imaging via stimulated emission depletion (STED) microscopy revealed filamentous RSV
Till F M Andlauer et al.
eLife, 3 (2014-11-14)
CIDE-N domains mediate interactions between the DNase Dff40/CAD and its inhibitor Dff45/ICAD. In this study, we report that the CIDE-N protein Drep-2 is a novel synaptic protein important for learning and behavioral adaptation. Drep-2 was found at synapses throughout the
Giuliana Fusco et al.
Nature communications, 7, 12563-12563 (2016-09-20)
α-synuclein (αS) is an intrinsically disordered protein whose fibrillar aggregates are the major constituents of Lewy bodies in Parkinson's disease. Although the specific function of αS is still unclear, a general consensus is forming that it has a key role
Anna Albisetti et al.
PLoS pathogens, 13(11), e1006710-e1006710 (2017-11-02)
Trypanosoma brucei belongs to a group of unicellular, flagellated parasites that are responsible for human African trypanosomiasis. An essential aspect of parasite pathogenicity is cytoskeleton remodelling, which occurs during the life cycle of the parasite and is accompanied by major
Philippe F Y Vincent et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(33), 10853-10869 (2014-08-15)
The hair cell ribbon synapses of the mammalian auditory and vestibular systems differ greatly in their anatomical organization and firing properties. Notably, vestibular Type I hair cells (VHC-I) are surrounded by a single calyx-type afferent terminal that receives input from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service