Skip to Content
Merck
All Photos(1)

Key Documents

40232

Sigma-Aldrich

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

≥97.0% (dried material, CHN)

Synonym(s):

3-(N,N-Dimethyldodecylammonio)propanesulfonate, 3-(N,N-Dimethyllaurylammonio)propanesulfonate, 3-(Dodecyldimethylammonio)propanesulfonate, 3-(Lauryldimethylammonio)propanesulfonate, Dodecyldimethyl(3-sulfopropyl)ammonium hydroxide inner salt, Lauryl sulfobetaine, SB3-12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
335.55
Beilstein:
4145308
EC Number:
MDL number:
UNSPSC Code:
12161900
eCl@ss:
32190102
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

description

zwitterionic

Assay

≥97.0% (dried material, CHN)

form

powder

mol wt

micellar avg mol wt 18,500

aggregation number

55

ign. residue

≤0.1%

loss

≤1% loss on drying, 110 °C

CMC

2-4 mM (20-25°C)

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤150 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

detection

SMILES string

CCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C17H37NO3S/c1-4-5-6-7-8-9-10-11-12-13-15-18(2,3)16-14-17-22(19,20)21/h4-17H2,1-3H3

InChI key

IZWSFJTYBVKZNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate has been used as a component of trypsin buffer for the dialysis of protein solution.

Other Notes

Zwitterionic surfactant which effectively solubilizes cardiac 5′-nucleotidase[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E H Chowdhury et al.
Journal of biotechnology, 119(4), 343-347 (2005-08-13)
Purification of plasmid DNA from bacteria is an essential tool in recombinant DNA technology and has become an essential task in laboratories to industries. Moreover, the recent progress of "Gene therapy" and "Genetic vaccination" also demands production of pharmaceutical grade
Masahiro Yoshizawa-Fujita et al.
The journal of physical chemistry. B, 114(49), 16373-16380 (2010-11-23)
We describe zwitterion, 3-(1-butyl-1H-imidazol-3-ium-3-yl)propane-1-sulfonate (Bimps), mixtures with 1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfoneamide (HN(Tf)(2)) as new proton transport electrolytes. We report proton transport mechanisms in the mixtures based on results from several methods including thermal analyses, the complex-impedance method, and the pulsed field gradient spin
Hongying Jia et al.
Journal of chromatography. A, 1143(1-2), 88-97 (2007-02-03)
With the accelerating development of new drugs, there is a high need for rapid and simple screening technologies. In this paper, a new in vitro method, dodecyl dimethyl betaine (BS-12) micellar liquid chromatography (MLC) was presented for prediction of protein-drug
Suguru Yamamoto et al.
Biochemistry, 43(34), 11075-11082 (2004-08-25)
In beta(2)-microglobulin-related (Abeta2M) amyloidosis, partial unfolding of beta(2)-microglobulin (beta2-m) is believed to be prerequisite to its assembly into Abeta2M amyloid fibrils in vivo. Although low pH or 2,2,2-trifluoroethanol at a low concentration has been reported to induce partial unfolding of
María Muñoz et al.
Langmuir : the ACS journal of surfaces and colloids, 20(25), 10858-10867 (2004-12-01)
Micellization in sodium dodecyl sulfate (SDS)-N-dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate and SDS-polyoxyethylenesorbitan monolaurate binary surfactant solutions was studied by means of conductivity and surface tension measurements. These studies showed that two types of micellar aggregates are present in the mixed micellar solutions. Two reactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service